Subtotal =  
Items =  
View Cart
Welcome
 
Home  >  Biosafety Cabinets (Class II)   

Class II B2 4-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-15)

YO-33515-15
Loading price...

Discontinued
Qty

  • Digital display provides constant feedback for uncompromised safety
  • Electronic motor uses 60% less energy than comparable models
  • Filter life graph reassures user that dangerous particles are not being released into the environment
  • Integrated lift knobs on the stainless steel work surface make cleaning easier
These cabinets have been designed to provide unprecedented assurance that you, your product, and your laboratory environment are protected. Typical applications include work with agents that require Biosafety Level 1, 2, or 3 containment and antineoplastic drug preparation. HEPA filter system removes 99.99% of 0.3-µm particles. Advanced filter monitoring system and ergonomic design increase your safety.


LCD safety monitoring system provides detailed information and warns when hazardous conditions are detected. Airflow alert warns you of a change in airflow, which could be caused by an obstruction and be potentially harmful, or when the filter has 20, 10, and 0% life remaining. Status line provides feedback on the system, including whether the system is “OK” or if the “sash is too high.” Continuous bar graph display provides information on the life of the HEPA filter and when it will need to be changed.


Electronically commutated motor (ECM) technology provides precisely controlled airflow with no need for additional airflow sensors. Motor is programmed after installation to deliver a consistent volume of air, even as the load on the HEPA filter changes, also providing longer filter life. ECM conserves energy by using up to 60% less than conventional motor systems. Quieter operation and less heat generation allow for more comfortable working conditions. ECM lasts approximately four times longer than conventional blower motors. Night-Smart system allows the unit to run at 30% normal speed when sash is completely closed to help maintain a clean environment when not in use; blower resumes normal operation when sash is raised.


Ergonomic design features include an angled sash for more comfortable viewing without glare, 29” of viewing height, bright fluorescent lighting for less eye strain, and a curved inlet grille to support your forearms. Large interior provides more cubic feet than any other biosafety cabinet available, and the leaktight 304 stainless steel liner includes a removable work surface with attached lift knobs for easier cleaning. All cabinets require a base stand, order separately.


Type A2 Biosafety Cabinets are designed to discharge HEPA-filtered air directly into the laboratory environment, while providing a nominal inflow velocity of 105 fpm and a downflow velocity of 55 fpm; maintaining approximately 70% air recirculation within the unit. A2 enclosures may also be vented to the outside, allowing you to use minute quantities of volatile toxic chemicals. Type A2 biosafety cabinets are available in 3-, 4-, 5-, and 6-ft lengths.


Type B2 Biosafety Cabinets are designed to be hard ducted to the outside and provide no recirculaton, with 100% of the air being exhausted. This feature allows these units to be used for work on agents treated with volatile toxic chemicals and radionuclides. Type B2 biosafety cabinets are available in 4- and 6-ft lengths. Blower is required, order separately.
What's Included:
cabinet, blower, light, and 8-ft power cord. Models with a 254-nm UV lamp are available.

Product Type Biosafety cabinets/enclosures
Interior dimensions (" W) 48.5
Interior dimensions (" H) 29.21875
Interior dimensions (" D) 17.59375
Exterior Dimensions (" W) 54.3125
Exterior Dimensions (" H) 61.40625
Exterior Dimensions (" D) 31.21875
Sash opening (in.) 8
Power (VAC) 115
Power (Hz) 60
Power (Amps) 15
Exhaust Volume (cfm) 743 - 771
Brand Labconco





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Related Products for Class II B2 4-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-15)
    Image
      Product Detail
    SKU

    Class II B2 4-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-35)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II A2 4-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-36)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II B2 6-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-55)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II B2 6-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-75)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II A2 4-ft Biosafety Cabinet with 10" sash opening and UV light (YO-33515-44)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II A2 3-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-04)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II A2 5-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-52)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II A2 5-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-60)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II A2 6-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-84)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Class II A2 4-ft Biosafety Cabinet with 8" sash opening and UV light 1 (YO-33515-28)

    Category: Biosafety Cabinets (Class II)
    Description: Labconco Purifier Logic Class II Biosafety Cabinets
    Brand: Labconco


    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews