Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-33859-03
Furnace,high Temp,1600c Box
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-33859-03
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Muffle Furnaces
Share
|
Furnace,high Temp,1600c Box (RHF16/3/3216P1)
YO-33859-03
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Combines high temperature capability with an advanced controller at a very affordable price
Heat to 1600°C in less than one hour
Program allows you to ramp temperature at controlled rates
This furnace’s high-temperature output is great for heat treating, reacting studies, sintering processes, and ceramic firing. Powerful silicon carbide heating elements ensure quick heating and a uniform chamber temperature. Quality chamber construction includes a durable refractory brick entrance, a long-life cast alumina hearth, and energy-efficient ceramic fiber insulation.
Engineered with your safety in mind, the furnace has a double shell construction to keep the outer case cool, a door switch that disengauges power when the door is opened, and the easy-to-operate vertical-swing door keeps heat away when opened. Furnace is equipped with a chimney vent for process exhaust.
Furnace temperature controller utilizes PID technology for highly accurate 16-segment programmability with up to 8 ramps (either up or down) and 8 dwells (time soaked). A three-color LCD display simultaneously indicates actual furnace temperature and set point temperature. Temperature may be displayed in °C or °F. Unique control system increases power to the heating elements to compensate for element resistance change during the aging process.
Specifications
Product Type
High-Temperature Furnace
Minimum temperature (º C)
0
Maximum temperature (º C)
1600
Minimum temperature (º F)
32
Maximum temperature (º F)
2912
Temp uniformity
±4°C
Chamber size (" H)
5.125
Chamber size (" D)
8
CE Compliance
Yes
Control type
PID microprocessor
Display
LCD
Manufacturer number
RHF16/3/3216P1
Power (VAC)
208/240
Power (Hz)
50/60
Model
RHF16/3/3216P1
Brand
Carbolite
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Furnace,high Temp,1600c Box (RHF16/3/3216P1)
Box Furnace
Temperature Box
High Temperature Box Furnace
Carbolite Muffle Furnaces
High Temperature Zirconia Sintering Muffle Furnace
Cole-Parmer Boxes
Cole Hdpe Boxes
Methanol 1 2 Gallon Storage Container
Cole-Parmer Accessories for all Glove Boxes
Trans-Boxes Plastic Storage Bins & Containers
Related Products for Furnace,high Temp,1600c Box (RHF16/3/3216P1)
Image
Product Detail
SKU
Carboliteheavy Duty Box Furnace (CWF13/13/3216P1)
Category: Muffle Furnaces
Description: Carbolite Heavy Duty Box Furnace
Brand:
Carbolite
Model Number:
CWF13/13/3216P1
YO-33859-02
More Details
Carboliteheavy Duty Box Furnace (CWF12/13/3216PI)
Category: Muffle Furnaces
Description: Carbolite Heavy Duty Box Furnace
Brand:
Carbolite
Model Number:
CWF12/13/3216PI
YO-33859-01
More Details
High-accuracy pressure transmitters with 3-ft cable, stripped ends; 1/ (YO-68971-01)
Category: High-Accuracy Pressure Transmitters
Description: Transmitters With 3-ft Cable
Brand:
Cole-Parmer
YO-68971-01
More Details
High-accuracy pressure transmitters with 3-ft cable, stripped ends; 1/ (2600BGF150233DB001)
Category: High-Accuracy Pressure Transmitters
Description: Transmitters With 3-ft Cable
Brand:
Cole-Parmer
Model Number:
2600BGF150233DB001
YO-68971-03
More Details
High-accuracy pressure transmitters with 3-ft cable, stripped ends; 1/ (2600BGF150233DB)
Category: High-Accuracy Pressure Transmitters
Description: Transmitters With 3-ft Cable
Brand:
Cole-Parmer
Model Number:
2600BGF150233DB
YO-68971-05
More Details
High-accuracy pressure transmitters with 3-ft cable, stripped ends; 1/ (2600BGF300233DB)
Category: High-Accuracy Pressure Transmitters
Description: Transmitters With 3-ft Cable
Brand:
Cole-Parmer
Model Number:
2600BGF300233DB
YO-68971-07
More Details
High-accuracy pressure transmitters with 3-ft cable, stripped ends; 1/ (2600BGF600233BD)
Category: High-Accuracy Pressure Transmitters
Description: Transmitters With 3-ft Cable
Brand:
Cole-Parmer
Model Number:
2600BGF600233BD
YO-68971-09
More Details
High-accuracy pressure transmitters with 3-ft cable, stripped ends; 1/ (2600BGG100233DB)
Category: High-Accuracy Pressure Transmitters
Description: Transmitters With 3-ft Cable
Brand:
Cole-Parmer
Model Number:
2600BGG100233DB
YO-68971-11
More Details
High-accuracy pressure transmitters with 3-ft cable, stripped ends; 1/ (2600BGG150233DB)
Category: High-Accuracy Pressure Transmitters
Description: Transmitters With 3-ft Cable
Brand:
Cole-Parmer
Model Number:
2600BGG150233DB
YO-68971-13
More Details
High-accuracy pressure transmitters with 3-ft cable, stripped ends; 1/ (2600BGG200233DB)
Category: High-Accuracy Pressure Transmitters
Description: Transmitters With 3-ft Cable
Brand:
Cole-Parmer
Model Number:
2600BGG200233DB
YO-68971-15
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Furnace,high Temp,1600c Box (RHF16/3/3216P1)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Furnace 8" tube length, 1300°C, 220-240VAC
|
Next: Furnaces with SSP control; 130 cu in; overall dimensions; 11 1/2" x 13 (9493300)
31125329-p-content_2 31125329pcontent2