Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-34106-00
Analtech CycloGraph
I TLC System
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-34106-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Thin Layer Chromatography (TLC)
Share
|
Analtech CycloGraph™ I TLC System (YO-34106-00)
YO-34106-00
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Put a new spin on preparative chromatography separate large-volume samples in 20 minutes or less
Simple fraction collection of up to four grams of sample material
Rotors are reusable up to 40 runs
CycloGraph enhances conventional TLC methods with centrifugal force. Perform your large-sample separations in 20 minutes – with no band scraping. TLC fractions spin-elute off the rotor edge into the radial trough and are funneled to your collection vessel. Reusable rotors are available with four adsorbent thicknesses. Save time and eliminate solvent transitions by alternating dried rotors. Pre-scraped rotors are ready for use straight from the box. Readily distinguish fraction bands using top-mounted UV light and rotors containing UV
254
indicator. An adjustable pump (0 to 15mL/min) precisely delivers solvent to inner rotor for continuous elution. Variable-speed motor and precision-made rotors produce optimal elution conditions and wobble-free operation.
What
Specifications
Brand
CycloGraph
Accessories
Qty
/
Price
Availability
Prescraped 2000 micron rotor for CycloGraph Chromatography System
YO-34106-22
EACH
Call for availability.
Add to Cart
Prescraped 4000 micron rotor for CycloGraph Chromatography System
YO-34106-24
EACH
Call for availability.
Add to Cart
Prescraped 6000 micron rotor for CycloGraph Chromatography System
YO-34106-26
EACH
Call for availability.
Add to Cart
Prescraped 8000 micron rotor for CycloGraph Chromatography System
YO-34106-28
EACH
Call for availability.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Common Chromatography Terms
Show Less...
Related Searches for Analtech CycloGraph™ I TLC System (YO-34106-00)
Cyclograph Thin Layer Chromatography (Tlc)
Chromatography Systems
Btx Electrocell Fusion System
Btx Electro Cell Fusion Systems
Fusion Life Science
Btx Life Science
Cole-Parmer Wilmad-Labglass Tlc Developing Cylinders
Cole-Parmer Rotary Evaporator Systems
Cole-Parmer Soxhlet Extraction Glassware Systems
Cole-Parmer Conical-Bottom Tank Systems
Related Products for Analtech CycloGraph™ I TLC System (YO-34106-00)
Image
Product Detail
SKU
Prescraped 2000 micron rotor for CycloGraph Chromatography System (YO-34106-22)
Category: Thin Layer Chromatography (TLC)
Description: Analtech CycloGraph I TLC System
Brand:
CycloGraph
YO-34106-22
More Details
Prescraped 4000 micron rotor for CycloGraph Chromatography System (YO-34106-24)
Category: Thin Layer Chromatography (TLC)
Description: Analtech CycloGraph I TLC System
Brand:
CycloGraph
YO-34106-24
More Details
Prescraped 6000 micron rotor for CycloGraph Chromatography System (YO-34106-26)
Category: Thin Layer Chromatography (TLC)
Description: Analtech CycloGraph I TLC System
Brand:
CycloGraph
YO-34106-26
More Details
Prescraped 8000 micron rotor for CycloGraph Chromatography System (YO-34106-28)
Category: Thin Layer Chromatography (TLC)
Description: Analtech CycloGraph I TLC System
Brand:
CycloGraph
YO-34106-28
More Details
Embryo manipulation system (YO-02894-00)
Category: Electroporation Systems
Description: BTX Electro Cell Fusion Systems
Brand:
BTX
YO-02894-00
More Details
Spacer Set for Cole-Parmer Vertical Slab Gel Systems; 14.5 cm Height, (YO-28573-00)
Category: Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Description: Cole-Parmer Vertical Slab Gel Spacer Set for Height-adjustable units
Brand:
Cole-Parmer
YO-28573-00
More Details
Cole-Parmer Gear Pump System, Benchtop Analog Drive, 0.092 mL/rev, 115 (YO-74013-00)
Category: Gear Pump Systems/Skids
Description: Cole-Parmer Benchtop Analog Gear Pump Drive Systems
Brand:
Cole-Parmer
YO-74013-00
More Details
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.092 mL/rev, 11 (YO-74014-00)
Category: Gear Pump Systems/Skids
Description: Cole-Parmer Benchtop Digital Gear Pump Drive Systems
Brand:
Cole-Parmer
YO-74014-00
More Details
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Category: Extraction Apparatus
Description: Soxhlet Extraction Systems
Brand:
Cole-Parmer
YO-99036-00
More Details
Fluidized sand bath systems; temperature range; 50-600°C; 117V; 2x (YO-01184-00)
Category: Waterless Bead/Sand Baths
Description: Fluidized Sand Bath Systems
Brand:
Techne
YO-01184-00
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Analtech CycloGraph™ I TLC System (YO-34106-00)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Analong input rate/totalizer, four SPDT relays and 4 to 20 mA output,
|
Next: Analytical Balance Dust Cover for Adam Equipment balances (8003)
31125329-p-content_2 31125329pcontent2