Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-34543-23
PYREX
®
Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL; tolerance, ±0.05mL
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-34543-23
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17104-00
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
$122.86 AUD/EACH/EACH
Usually ships in 23 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Glass and Plastic Burettes
Share
|
PYREX Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL
YO-34543-23
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Colored scale, Class A, PTFE
®
stopcock plug
The capacity tolerance on these burettes is established by ASTM E-287. The burettes are calibrated in accordance with ASTM E-542. Tips are carefully drawn from accurate bore tubing to ensure proper drainage rates. Burettes have colored markings, fine sharp lines, and large, easy-to-read numbers. The stopcock barrel has a smooth micro-finish to assure leak-resistant performance. Each burette is supplied with a dust cover, and all sizes are furnished with a 2-mm bore stopcock plug.
Specifications
Product Type
Class A Burettes
Subdivisions (mL)
0.1
Tolerance
±0.05 mL
Capacity (mL)
50
Brand
Pyrex
Manufacturer number
2103-50
Model
2103-50
Accessories
Qty
/
Price
Availability
Burette brush, 15"L x 3/16" diameter, pack of 12
YO-84550-60
PACKAGE of 12
Call for availability.
Add to Cart
Burette brush, 16-1/2
YO-84550-61
PACKAGE of 12
Call for availability.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
YO-17104-00
EACH
Usually ships in 23 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related Searches for PYREX Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL
Pyrex Brand 2103 Class a Burettes
50 Ml Graduated Cylinder
50 Ml Burette Farinograph
50 Ml Automatic Burette
50 Ml Burette Stopcocks Bellwether
50 Ml Pyrex Flask
10 Ml Graduated Burettes
Centrifuges Capacity 10 Ml
Burette Glassware 10 Ml 27
Pyrex Brand 5642 Glass Volumetric Flasks
Related Products for PYREX Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL
Image
Product Detail
SKU
PYREX Brand 2103 Burette; capacity, 10 mL; graduation interval, 0.05mL
Category: Glass and Plastic Burettes
Description: PYREX Brand 2103 Class A Burettes
Brand:
Pyrex
Model Number:
2103-10
YO-34543-21
More Details
PYREX Brand 2103 Burette; capacity, 25 mL; graduation interval, 0.10mL
Category: Glass and Plastic Burettes
Description: PYREX Brand 2103 Class A Burettes
Brand:
Pyrex
Model Number:
2103-25
YO-34543-22
More Details
Pyrex Brand 2110 Burette; capacity, 50 mL; graduation interval, 0.10m
Category: Glass and Plastic Burettes
Description: Pyrex Brand 2110 Burettes
Brand:
Pyrex
Model Number:
2110-50
YO-34544-22
More Details
PYREX Brand 2103 Burette; capacity,100 mL; graduation interval, 0.20mL
Category: Glass and Plastic Burettes
Description: PYREX Brand 2103 Class A Burettes
Brand:
Pyrex
Model Number:
2103-100
YO-34543-24
More Details
PYREX Brand 5640 Volumetric Flask; 50 mL, pack of 6
Category: Glass Volumetric Flasks
Description: PYREX Brand 5640 Glass Volumetric Flasks
Brand:
Pyrex
YO-34505-06
More Details
PYREX Brand 5640 Volumetric Flask; 50 mL, case of 12
Category: Glass Volumetric Flasks
Description: PYREX Brand 5640 Glass Volumetric Flasks
Brand:
Pyrex
YO-34505-26
More Details
PYREX Brand 5580 Volumetric Flask; 50 mL, pack of 6
Category: Glass Volumetric Flasks
Description: PYREX Brand 5580 Glass Volumetric Flasks
Brand:
Pyrex
YO-34560-06
More Details
PYREX Brand 5580 Volumetric Flask; 50 mL, case of 12
Category: Glass Volumetric Flasks
Description: PYREX Brand 5580 Glass Volumetric Flasks
Brand:
Pyrex
YO-34560-26
More Details
PYREX Brand 5642 Volumetric Flask; 50 mL, pack of 6
Category: Glass Volumetric Flasks
Description: PYREX Brand 5642 Glass Volumetric Flasks
Brand:
Pyrex
YO-34561-05
More Details
PYREX Brand 5642 Volumetric Flask; 50 mL, case of 12
Category: Glass Volumetric Flasks
Description: PYREX Brand 5642 Glass Volumetric Flasks
Brand:
Pyrex
YO-34561-25
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
PYREX Brand 2103 Burette; capacity, 50 mL; graduation interval, 0.10mL
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: PYREX Brand 2103 Burette; capacity, 25 mL; graduation interval, 0.10mL
|
Next: PYREX Brand 2103 Burette; capacity,100 mL; graduation interval, 0.20mL
31125329-p-content_2 31125329pcontent2