Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
PYREX Brand 4320 round bottom flask 5000 mL pack of 1
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-34553-07
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Single Neck Flasks
Share
|
PYREX Brand 4320 round-bottom flask; 5000 mL, pack of 1
YO-34553-07
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Boiling, round bottom, short neck, joint
PYREX
®
brand boiling flasks are carefully designed and manufactured from borosilicate glass. Uniform wall thicknesses, a characteristic of all round-bottom, PYREX glassware,provides a balance between expansion and mechanical strength for greater shock resistance and versatility—use boiling flasks for a variety of mixing, heating and boiling applications commonly encountered in most laboratory work. The normal working temperature is 230degC.
Specifications
Capacity (mL)
5000
Material
Glass
Height (cm)
30.00
Quantity
1
Brand
Pyrex
Manufacturer number
4320-5L
Accessories
Qty
/
Price
Availability
PTFE Boiling Stones
YO-01199-00
POUND
Call for availability.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related Searches for PYREX Brand 4320 round-bottom flask; 5000 mL, pack of 1
Three Neck Round Bottom Flask
5000 Ml Glass Round Bottom Flasks
5000 Ml Round Bottom Flask
1x 1500 Ml Round Bottom Flask
6000 Ml Round Bottom Flask
Round Bottom Flask Magnetic Stirrer
Manifold Lyophilizer Round Bottom Flasks
1000 Ml Round Bottom Flask
Pyrex Round Bottom Flask
250 Ml Round Bottom Flask
Related Products for PYREX Brand 4320 round-bottom flask; 5000 mL, pack of 1
Image
Product Detail
SKU
PYREX Brand 4320 round-bottom flask; 1000 mL, pack of 1
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-05
More Details
PYREX Brand 4320 round-bottom flask; 2000 mL, pack of 1
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-06
More Details
PYREX Brand 4320 round-bottom flask; 5000 mL, case of 4
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-27
More Details
PYREX Brand 4320 round-bottom flask; 100 mL, pack of 2
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-02
More Details
PYREX Brand 4320 round-bottom flask; 250 mL, pack of 2
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-03
More Details
PYREX Brand 4320 round-bottom flask; 500 mL, pack of 2
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-04
More Details
PYREX Brand 4320 round-bottom flask; 12000 mL, case of 1
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-28
More Details
PYREX Brand 4320 round-bottom flask; 50 mL, pack of 2
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-01
More Details
PYREX Brand 4320 round-bottom flask; 50 mL, case of 12
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-21
More Details
PYREX Brand 4320 round-bottom flask; 100 mL, case of 12
Category: Single Neck Flasks
Description: PYREX Brand 4320 Glass, Round Bottom Flasks
Brand:
Pyrex
YO-34553-22
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
PYREX Brand 4320 round-bottom flask; 5000 mL, pack of 1
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: PYREX Brand 4320 round-bottom flask; 5000 mL, case of 4
|
Next: PYREX Brand 4980 flask; 10 mL, case of 12
31125329-p-content_2 31125329pcontent2