Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Corning milk dilution bottle wide mouth non graduated 160 mL Pack of 12
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-34583-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Dilution Bottles
Share
|
Corning® milk dilution bottle; wide mouth, non-graduated, 160 mL. (1368-160) (YO-34583-00)
YO-34583-00
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Glass bottles meet the requirements for milk dilution bottles stated in the Standard Methods for the Examination of Dairy Products, published by the American Public Health Association. Graduated bottles have a cut-line graduation mark at 99 ± 1mL. Bottles are autoclavable and phenolic cap with rubber liner withstands wet sterilization up to 121C. Choose narrow mouth bottles for general storage of solutions and microbiological work. Wide mouth bottles make pouring and measuring liquids easier, and simplify the addition of dry solids. Bottles have a 160 mL capacity and measure 150 mm H x 44 mm W x 44 mm D.
Specifications
Product Type
Dilution bottles
Material
Glass
Capacity (Liters)
0.16
Capacity (mL)
160
Qty/pk
12
Dimensions
150 mm H x 44 mm W x 44 mm D
Brand
Corning
Model
1368-160
Manufacturer number
1368-160
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
11-12 International Catalog
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Cole-Parmer Introduces Enhancements to Life Science Offering
Show Less...
Related Searches for Corning milk dilution bottle; wide mouth, non-graduated, 160 mL. Pack
Autoclavable Milk Dilution Bottles
Corning Glass Milk Dilution Bottles
Wide Mouth Milk Dilution Bottles
250 Ml Milk Dilution Bottles
Nalgene Economy Bottle Wide Mouth Ppco
Nalgene Economy Bottle Wide Mouth Hdpe
Nalgene Passport Ip2 Bottle Wide Mouth
Dilution Bottles 200 Ml
Microbiology Dilution Bottles
Thermo Scientific Wide-Mouth Unitary Wash Bottles
Related Products for Corning milk dilution bottle; wide mouth, non-graduated, 160 mL. Pack
Image
Product Detail
SKU
Corning milk dilution bottle; wide mouth, non-graduated, 160 mL. Case
Category: Dilution Bottles
Description: CORNING Milk Dilution Bottles
Brand:
Corning
Model Number:
1368-160
YO-34583-20
More Details
Corning milk dilution bottle; narrow mouth, non-graduated, 160 mL. Pac
Category: Dilution Bottles
Description: CORNING Milk Dilution Bottles
Brand:
Corning
Model Number:
1367-160
YO-34582-00
More Details
Corning milk dilution bottle; narrow mouth, non-graduated, 160 mL. Cas
Category: Dilution Bottles
Description: CORNING Milk Dilution Bottles
Brand:
Corning
Model Number:
1367-160
YO-34582-20
More Details
Corning milk dilution bottle; wide mouth, graduated, 160 mL. Pack of 1
Category: Dilution Bottles
Description: CORNING Milk Dilution Bottles
Brand:
Corning
Model Number:
1373-160
YO-34585-00
More Details
Corning milk dilution bottle; wide mouth, graduated, 160 mL. Case of 4
Category: Dilution Bottles
Description: CORNING Milk Dilution Bottles
Brand:
Corning
Model Number:
1373-160
YO-34585-20
More Details
Corning milk dilution bottle; narrow mouth, graduated, 160 mL. Pack of
Category: Dilution Bottles
Description: CORNING Milk Dilution Bottles
Brand:
Corning
Model Number:
1372-160
YO-34584-00
More Details
Corning milk dilution bottle; narrow mouth, graduated, 160 mL. Case of
Category: Dilution Bottles
Description: CORNING Milk Dilution Bottles
Brand:
Corning
Model Number:
1372-160
YO-34584-20
More Details
Thermo Scientific Nalgene Bottle, Wide Mouth, FEP 125mL, 1/pk
Category: Wide-Mouth Plastic Bottles
Description: Thermo Scientific Nalgene Wide-Mouth Flourinated Ethylene Propylene Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2100-0004
YO-06044-10
More Details
Thermo Scientific Nalgene Bottle, Wide Mouth, FEP 250mL, 1/pk
Category: Wide-Mouth Plastic Bottles
Description: Thermo Scientific Nalgene Wide-Mouth Flourinated Ethylene Propylene Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2100-0008
YO-06044-20
More Details
Thermo Scientific Nalgene Bottle, Wide Mouth, FEP 500mL, 1/pk
Category: Wide-Mouth Plastic Bottles
Description: Thermo Scientific Nalgene Wide-Mouth Flourinated Ethylene Propylene Bottles
Brand:
Thermo Scientific Nalgene
Model Number:
2100-0016
YO-06044-30
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Corning milk dilution bottle; wide mouth, non-graduated, 160 mL. Pack
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Corning milk dilution bottle; wide mouth, non-graduated, 160 mL. Case
|
Next: Corning Nine-Position Stirrer, 120 VAC, 60 Hz
31125329-p-content_2 31125329pcontent2