Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
304 Stainless Steel Tubing 1 8 OD 0 085 ID 50ft roll
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-34680-50
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Accessories for Gas Chromatography (GC)
Share
|
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
YO-34680-50
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
304 Stainless Steel tubing is perfect for use in gas chromatography
Make your own GC columns; simply cut the tubing to length and coil to fit your gas chromatograph. Special cleaning process leaves tubing free of residual materials. Maximum pressure of 10,000 psig. Fittings are not inlcuded. Tubing comes in 50 foot lengths.
Specifications
Material
304 Stainless Steel
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Sourcing Hydrogen for Gas Chromatography
Chromatography Column and Phase Cross-Reference
Common Chromatography Terms
Show All...
Show Less...
Related Searches for 304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Coiled Stainless Steel Tubing 1 8
304 Stainless Steel Tubing Suppliers
304 Stainless Steel Tubing Coils
304 Stainless Steel Tubing Od Tolerance
304 Stainless Steel Tubing
304 Stainless Steel Tubing 25 Gauge
304 Stainless Steel Tubing Vacuum
Stainless Steel Tube 1 4 08
304 Stainless Steel Tubing 8 Ft
Rubber Vacuum Tubing 1 8 Od
Related Products for 304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
Image
Product Detail
SKU
304 Stainless Steel Tubing, 1/4" OD, 0.210" ID, 50ft roll (YO-34680-60)
Category: Accessories for Gas Chromatography (GC)
Description: Stainless Steel Tubing
Brand:
Cole-Parmer
YO-34680-60
More Details
304 Stainless Steel manifold; 8 outlets; 1/4" inlet NPT(F) (YO-31521-49)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-49
More Details
304 Stainless Steel manifold; 8 outlets; 1/2" inlet NPT(F) (YO-31521-53)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-53
More Details
304 Stainless Steel manifold; 2 outlets; 1/4" inlet NPT(F) (YO-31521-31)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-31
More Details
304 Stainless Steel manifold; 2 outlets; 3/8" inlet NPT(F) (YO-31521-33)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-33
More Details
304 Stainless Steel manifold; 2 outlets; 1/2" inlet NPT(F) (YO-31521-35)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-35
More Details
304 Stainless Steel manifold; 4 outlets; 1/4" inlet NPT(F) (YO-31521-37)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-37
More Details
304 Stainless Steel manifold; 4 outlets; 3/8" inlet NPT(F) (YO-31521-39)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-39
More Details
304 Stainless Steel manifold; 4 outlets; 1/2" inlet NPT(F) (YO-31521-41)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-41
More Details
304 Stainless Steel manifold; 6 outlets; 1/4" inlet NPT(F) (YO-31521-43)
Category: Metal and Thermoplastic Manifolds
Description: 304 Stainless Steel Manifolds
Brand:
Cole-Parmer
YO-31521-43
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
304 Stainless Steel Tubing, 1/8" OD, 0.085" ID, 50ft roll (YO-34680-50)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: 304 Stainless Steel Tubing, 1/4" OD, 0.210" ID, 50ft roll (YO-34680-60)
|
Next: 304 STAINLESS STEEL WORK SURFACE 48IN WIDE BY 35.5IN DEEP (YO-28614-70)
31125329-p-content_2 31125329pcontent2