Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-74000-72
Representative Photo Only. Sandpiper® Air-Powered Double-Diaphragm Polypropylene Pump, Polypropylene, 4 GPM, PTFE Diaphragm
Click to enlarge
More Product Details
Specifications
Technical Resources
Flow Charts
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-74000-72
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Air-Powered Double-Diaphragm Pumps
Share
|
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (PB.25,TT3PP.)
YO-74000-72
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Move fluids with viscosities up to 11,000 cp
Air-powered motor eliminates sparking concerns associate with electrical pumps
Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing
Non-metallic pumps are ideally suited for highly corrosive fluids intermittent/on-demand, portable low abrasive fluids, and suspended solids
Safely handles abrasive fluids and slurriesideal for high-viscosity and shear sensitive materials
Encapsulated muffler provides excellent sound dampening
These pumps can move high-viscosity fluids such as paint, sludge, and sewage. However, flow rate and pressure depends on the application. If you have questions about the compatibility of your high-viscosity fluid, call our Application Specialists for more information.
Order air filters/regulators to remove impurities from your air line and to help regulate working pressure. Combination air filter/regulators feature polypropylene filter, 0- to 160-psi gauge, and a polycarbonate bowl. Temperature range is 0 to 125°F (17 to 51°C).
Specifications
Product Type
Air-Powered Diaphragm Pumps
Wetted materials
Polypropylene, PVDF, and conductive acetal with TPE (thermal plastic elastomers) and PTFE options in diaphragms and check valves
Port size
1/4" NPT(F)
Max flow rate (LPM)
15.1
Max flow rate (GPM)
4
Max viscosity (cp)
11,000
Max pressure
100 psi
Max temperature (º C)
66
Max temperature (º F)
150
Run dry
Yes
Suction lift (wet)
10 ft
Air inlet (SCFM)
1 to 6 scfm
Air inlet (connection)
1/4" NPT(F)
Air inlet (max psi)
100
Duty cycle
Continuous
Max solids dia.
1/32"
Dimensions
7"L x 5-1/2"W x 7-13/16"H
Brand
Sandpiper
Manufacturer number
PB 1/4,TT3PP.
Model
PB.25,TT3PP.
Technical Resources
Flow curve for 74000-72
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Positive Displacement Pump Types
How to Read a Pump Curve
Guide to Liquid Pumps
Show All...
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Related Searches for Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (PB.25,TT3PP.)
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Sandpiper Air-Powered Double-Diaphragm Pvdf Pumps
Sandpiper Air-Powered Double-Diaphragm Pumps
Sandpiper Air-Powered Double-Diaphragm Stainless Steel Pumps
Polypropylene Air-Powered Double-Diaphragm Pumps
Aro Air-Powered Double-Diaphragm Pumps
Santoprene Air-Powered Double-Diaphragm Pumps
Kynar Air-Powered Double-Diaphragm Pumps
Ptfe Air-Operated Double-Diaphragm Drum Pumps
Kynar Air-Operated Double-Diaphragm Drum Pumps
Related Products for Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (PB.25,TT3PP.)
Image
Product Detail
SKU
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (S05B2P1TPNS000)
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Brand:
Sandpiper
Model Number:
S05B2P1TPNS000
YO-74000-00
More Details
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (S1FB3P1PPUS100)
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Brand:
Sandpiper
Model Number:
S1FB3P1PPUS100
YO-74000-02
More Details
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (S07B1P1PPNS000.)
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Brand:
Sandpiper
Model Number:
S07B1P1PPNS000.
YO-74000-11
More Details
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (S1FB3P2PPUS100)
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Brand:
Sandpiper
Model Number:
S1FB3P2PPUS100
YO-74000-12
More Details
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyl
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Brand:
Sandpiper
Model Number:
S07B1P2PPNS000.
YO-74000-13
More Details
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (SO5B2P2TPNS000)
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Brand:
Sandpiper
Model Number:
SO5B2P2TPNS000
YO-74000-42
More Details
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (PB.25,TS3PP.)
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Brand:
Sandpiper
Model Number:
PB.25,TS3PP.
YO-74000-70
More Details
Sandpiper Air-Powered Double-Diaphragm PVDF Pump, PVDF, 23 GPM, Santop
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm PVDF Pumps
Brand:
Sandpiper
Model Number:
S07B1K1KPNS000.
YO-74000-21
More Details
Sandpiper Air-Powered Double-Diaphragm PVDF Pump, PVDF, 45 GPM, Santop
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm PVDF Pumps
Brand:
Sandpiper
Model Number:
S1FB3K1KPUS000
YO-74000-22
More Details
Sandpiper Air-Powered Double-Diaphragm PVDF Pump, PVDF, 23 GPM, PTFE D
Category: Air-Powered Double-Diaphragm Pumps
Description: Sandpiper Air-Powered Double-Diaphragm PVDF Pumps
Brand:
Sandpiper
Model Number:
S07B1K2KQNS000.
YO-74000-23
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (PB.25,TT3PP.)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (PB.25,TS3PP.)
|
Next: Sandpiper Air-Powered Double-Diaphragm Polypropylene Pump, Polypropyle (S05B2P1TPNS000)
31125329-p-content_2 31125329pcontent2