Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-35423-10
Oakton® Waterproof EcoTestr™ pH 2 Pocket pH Tester
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-35423-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17101-45
NIST Traceable Calibration; pH Tester and Pens
$121.00 AUD/EACH/EACH
Usually ships in 23 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Oakton EcoTestr Pocket Testers
Share
|
Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
YO-35423-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Easy-to-read display with an easy-to-afford price!
Ergonomically designed LCD means measurements can be seen at a glance
Economical and easy to use
Waterproof and dustproof housing designed for use in dirty and damp field conditions
Simply dip and read the orientation of the display means no need to turn your head or tilt the tester. This low-cost meter is perfect for most applications. The single-junction electrode is recessed to protect against breakage. The unit offers up to three-point calibration with auto-buffer recognition for quick calibrations, automatic temperature compensation for accurate reading under changing conditions, hold function to lock measure value, and auto-off to prolong battery life. Built-in belt clip allows you to hang your meter when not in use.
What's Included:
four 1.5 V button cell batteries.
Specifications
Product Type
pH Tester
Meter style
Tester
Dimensions (" W)
1-5/8
Dimensions (" H)
6-1/4
Dimensions (" D)
1-1/8
Range (pH)
0.00 to 14.00
Range (Temperature °C)
5 to 50
Range (Temperature °F)
23 to 221
Resolution (pH)
0.1
Resolution (Temperature)
0.1
Accuracy (pH)
±0.1 pH
Temperature compensation
Automatic
Display
LCD
Calibration
Up to 3 points
Waterproof
Yes
Buffer recognition
USA, NIST
Power
Four 1.5 V batteries (included)
CE Compliance
Yes
Brand
Oakton
Model
EcoTestr pH 2
Accessories
Qty
/
Price
Availability
Oakton buffer pack, 500-ml each of 4.01, 7.00, and 10.00 buffer
YO-05942-10
PACKAGE of 3
Call for availability.
Add to Cart
Batteries, 1.5V button cells, 6/PK
YO-09377-16
PACKAGE of 6
Call for availability.
Add to Cart
Deluxe calibration kit
YO-35624-70
EACH
Call for availability.
Add to Cart
Oakton pH Buffer Pouches, Assortment
YO-35653-04
PACKAGE of 20
Call for availability.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Calibration; pH Tester and Pens
YO-17101-45
EACH
Usually ships in 23 days.
Add to Cart
Technical Resources
Instruction Manual
Oakton EcoTestr pH 2 (609 KB)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
More info
Oakton Catalogo en Espanol.pdf (8.3 MB)
Related technical articles (Roll over for summary)
Le secret pour assurer le bon fonctionnement d'un pH-mètre? Les électrodes!
The Secret to Well-Functioning pH Meters? (It's the Electrodes!)
Double Junction pH Tester-The Next Generation of Long Life Pocket-Sized pH Meters
Show All...
pH FAQs
Testing the pH Value of Pure Water
Testing the pH value of Meat
Testing the pH in Yogurt Production
Testing the pH value of Wine
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
pH Electrode Theory
Oakton® pH meter becomes a TV star!
Measuring the pH of Pure Water
pH Systems
pH Temperature Compensation
Temperature Compensation for pH Instruments
pH Measurement FAQs
Show Less...
Related Searches for Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
Italian Pocket Ph Meter
B 211 212 Pocket Ph Meter
Oakton Ecotestr Ph 2 Tester
Oakton Ecotestr Ph 2
Pocket Ph Meter with Isfet Sensor
Portable Pocket Ph Meter
Pocket Ph Meter Soil
2 Pocket
Oakton Waterproof Ph Tester
Oakton Waterproof Ph/Con 10 Ph/Conductivity Meters
Related Products for Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
Image
Product Detail
SKU
Oakton PTTestr 35 Waterproof Multiparameter pH/TDS pocket tester
Category: Multiparameter Pocket Testers
Description: Oakton Waterproof PTTestr 35 Pocket Multiparameter Testers
Brand:
Oakton
Model Number:
PTTestr 35
YO-35425-05
More Details
Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
Brand:
Oakton
Model Number:
pHTestr
10 BNC
YO-35634-14
More Details
Oakton EcoTestr TDS Low pocket TDS tester
Category: Oakton EcoTestr Pocket Testers
Description: Oakton EcoTestr TDS Low Pocket Tester
Brand:
Oakton
Model Number:
EcoTestr TDS Low
YO-35462-10
More Details
Oakton EcoTestr TDS High pocket TDS tester
Category: Oakton EcoTestr Pocket Testers
Description: Oakton EcoTestr TDS High Pocket Tester
Brand:
Oakton
Model Number:
EcoTestr TDS High
YO-35462-15
More Details
Oakton EcoTestr EC Low pocket conductivity tester
Category: Oakton EcoTestr Pocket Testers
Description: Oakton EcoTestr EC Low Pocket Tester
Brand:
Oakton
Model Number:
EcoTestr EC Low
YO-35462-30
More Details
Oakton EcoTestr EC High pocket conductivity tester
Category: Oakton EcoTestr Pocket Testers
Description: Oakton EcoTestr EC High Pocket Tester
Brand:
Oakton
Model Number:
EcoTestr EC High
YO-35462-35
More Details
Oakton PCTestr 35 Waterproof Multiparameter pH/conductivity pocket tes
Category: Multiparameter Pocket Testers
Description: Oakton Waterproof PCTestr 35 Pocket Multiparameter Testers
Brand:
Oakton
Model Number:
PCTestr 35
YO-35425-00
More Details
Oakton Waterproof ORPTestr 10 Pocket ORP Tester
Category: pH/Ion Pocket Testers
Description: Oakton Waterproof ORPTestr 10 Pocket ORP Tester
Brand:
Oakton
Model Number:
ORPTestr
10
YO-35650-10
More Details
Oakton Waterproof ORP Testr 10 BNC Pocket ORP Tester
Category: Pocket Testers
Description: Oakton Waterproof ORPTestr 10 BNC Pocket ORP Tester
Brand:
Oakton
Model Number:
ORPTestr
10 BNC
YO-35650-14
More Details
Hach waterproof ISFET pH/Ion meter only
Category: pH/Ion Pocket Testers
Description: Waterproof ISFET Handheld pH/mV/Ion Meters
Brand:
Hach
Model Number:
IQ160GNP
YO-99361-04
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Oakton waterproof DO meter replacement probe with 25-ft cable
|
Next: Oakton Waterproof ORP Testr 10 BNC Pocket ORP Tester
31125329-p-content_2 31125329pcontent2