Oakton Waterproof PCD 650 Multiparameter Meter Only
YO-35434-02
Loading price...
Call for availability.
Qty
  • Expanded range, resolution, and accuracy, plus increased memory and advanced communications!
  • No more excuses for missing data! Out-of-date or unperformed calibrations are now things of the past!
  • Don’t waste time looking for those cables!
  • Use in dirty and wet environments
This top-of the-line waterproof meter offers features that are typically found on much more expensive models. The IP67-rated housing allows use in dirty and wet environments without harm to the meter. Datalog up to 500 points with time-and-date stamp. User-selectable “calibration due” alarms, automatic or manual standards recognition, and single or multiple calibration points provide flexibility in meeting your data quality objectives. Infrared (IrDA) wireless technology lets you easily download data—no need for additional cords. The large backlit LCD shows the measured value, measured units, temperature, time, battery life, and menu options.
The pH range is from –2.000 to 20.000 with accuracies up to ±0.002. The pH calibration capabilities include up to six calibration points, a user-settable “calibration due” alarm, and up to 15 buffer options with auto-buffer recognition of USA, NIST, DIN, and PWB standards.
Meter measures conductivity (up to 500 mS), TDS, salinity, or resistivity with either a 2-or 4-electrode conductivity cell. Linear or pure-water temperature compensation curve provides accurate data at all levels of conductivity. Also features set point alarms to indicate when concentrations are outside of programmed limits.
Meter determines oxygen saturation up to 600% and concentration up to 90 mg/L. The galvanic dissolved oxygen probe does not require any warm-up time so you can take readings immediately. Salinity correction is automatically compensated for after manual input of concentration. Barometric pressure is automatically compensated for by the included barometer.
What's Included:
data management software, probe holder, and batteries.
Meter with probes adds: double-junction pH electrode, 2-cell conductivity cell, and a dissolved oxygen probe.
Meter kit further adds: pH calibration buffers, conductivity standards, dissolved oxygen probe replacement membrane kit, protective rubber boot, and hard carrying case.
Meter style Portable
Temperature compensation Automatic or manual
Display LCD
CE Compliance Yes
Accessories
Qty/Price
Availability
EACH
Call for availability.
EACH
Usually ships in 14 days or sooner.
EACH
Call for availability.
EACH
Call for availability.
EACH
Call for availability.
EACH
Call for availability.
Adapter 110/220 VAC
YO-35418-83
EACH
Usually ships in 14 days or sooner.
EACH
Call for availability.
EACH
Usually ships in 14 days or sooner.
EACH
Usually ships in 14 days or sooner.
EACH
Call for availability.
EACH
Call for availability.
EACH
Usually ships in 14 days or sooner.
EACH
Usually ships in 14 days or sooner.
EACH
Usually ships in 14 days or sooner.
EACH
Usually ships in 14 days or sooner.
EACH
Usually ships in 14 days or sooner.
RS-232 Interface
YO-91100-85
EACH
Usually ships in 14 days or sooner.
Calibrations
Qty/Price
Availability
EACH
Usually ships in 25 days.
EACH
Usually ships in 21 days.
Technical Resources
Related Products for Oakton Waterproof PCD 650 Multiparameter Meter Only
Image
  Product Detail
SKU
Oakton Waterproof PCD 650 Multiparameter Meter Kit
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PCD 650 Multiparameter Meter
Brand: Oakton
Oakton PCD 650 Waterproof Multiparameter Meter Kit w/ NIST Cal Cert
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PCD 650 Multiparameter Meter
Brand: Oakton
Oakton Waterproof PCD 650 Multiparameter Meter and Probes
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PCD 650 Multiparameter Meter
Brand: Oakton
Oakton PCD 650 Waterproof Multiparameter Meter w/Probes and NIST Cal Cert
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PCD 650 Multiparameter Meter
Brand: Oakton
Oakton PCD 650 Waterproof Multiparameter Meter w/ NIST Cal Cert
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PCD 650 Multiparameter Meter
Brand: Oakton
Oakton Waterproof PC 650 Meter with probes
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PC 650 pH/Conductivity Meters
Brand: Oakton
Oakton Waterproof PD 650 Meter with probes with NIST-traceable Calibration
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PD 650 pH/Dissolved Oxygen Meters
Brand: Oakton
Oakton Waterproof PD 650 Meter only with NIST-traceable Calibration
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PD 650 pH/Dissolved Oxygen Meters
Brand: Oakton
Oakton Waterproof PD 650 Meter Kit witn NIST-traceable Calibration
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PD 650 pH/Dissolved Oxygen Meters
Brand: Oakton
Oakton Waterproof PC 650 Meter with probes and NIST-traceable Calibration
Category: Handheld Multiparameter Meters
Description: Oakton Waterproof PC 650 pH/Conductivity Meters
Brand: Oakton
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call