Oakton 2-in-1 Waterproof Infrared Thermometer (7199OK)

YO-35625-41
Loading price...

Usually ships in 14 days or sooner.
Qty

  • A thermometer for all applications
  • Go/no-go LEDs provide quick check of HACCP food zones
  • IP65 rated waterproof housing
Ultra thin 2-in-1 waterproof contact/noncontact thermometer features a HACCP food safety LED warning. LED’s rapidly indicate if food temperature is within a safe or unsafe range. Green light indicate safe temperatures below 40°F (4°C) and above 140°F (60°C). Red light indicates the HACCP danger zone of 40 to 140°F (4 to 60°C). The built-in stainless steel type-K thermocouple probe swings out for internal temperature checks, then folds in for storage. IR mode provides quick, noncontact surface measurements. Bright white LED pointer illuminates the target measurement area and is ideal for close working distances of 2 to 12”. Product also features adjustable emissivity, Celsius or Fahrenheit selectable, min/max, auto data hold, lock
Product Type Infrared Thermometer
Distance to target size ratio 2.5:1
Min temperature (º C) -55
Max temperature (º C) 250
Min temperature (º F) -67
Max temperature (º F) 482
Accuracy (º C) IR: ±0.1°C from -33 to 0°C, ±1°C from 0 to 65°C, ±1.5% of reading from 65 to 300°C, Probe: ±0.5°C from -9 to 100°C else ±0.9% of reading
Accuracy (º F) IR: ±0.1°F from -27 to 32°F, ±1°F from 32 to 149°F, ±1.5% of reading from 149 to 572°F, Probe: ±0.5°F from 15 to 212°F else ±0.9% of reading
Emissivity Adjustable
Sighting type LED (non-laser)
Power two AAA batteries (included)
Display 4-digit LCD, 0.75 in W x 1 in H
Low battery indicator Yes
CE Compliance Yes
Manufacturer number 7199OK
Brand Oakton

Accessories
Qty/Price
Availability

PACKAGE of 12

Usually ships in 14 days or sooner.

BOX of 100

Usually ships in 14 days or sooner.
Calibrations
Qty/Price
Availability

EACH
Usually ships in 19 days.




Technical Resources
 
Related Products for Oakton 2-in-1 Waterproof Infrared Thermometer (7199OK)
Image
  Product Detail
SKU

Oakton Food TempTestr Mark-I Infrared Thermometer

Category: Food Infrared Thermometers
Description: Oakton Food TempTestr Basic Infrared Thermometer
Brand: Oakton


Oakton Food TempTestr Mark-I Infrared Thermometer Kit

Category: Food Infrared Thermometers
Description: Oakton Food TempTestr Basic Infrared Thermometer
Brand: Oakton


Oakton Food TempTestr Mark-I Infrared Thermometer with NIST-traceable Calibration

Category: Food Infrared Thermometers
Description: Oakton Food TempTestr Basic Infrared Thermometer
Brand: Oakton


4-Carbethoxy-2-ethyl-3-methyl-2-cyclohexen-1-one tech. (25g) (AC155680250)

Category: Camphan to Carbowa
Description: 4-Carbethoxy-2-ethyl-3-methyl-2-cyclohexen-1-one
Brand: Cole-Parmer


D(+)-2-Amino-1-butanol 98+% (ca 95% e.e.) (1g) (AC158080010)

Category: Ametho to Aminob
Description: D(+)-2-Amino-1-butanol
Brand: Cole-Parmer


D(+)-2-Amino-1-butanol 98+% (ca 95% e.e.) (2.5g) (AC158080025)

Category: Ametho to Aminob
Description: D(+)-2-Amino-1-butanol
Brand: Cole-Parmer


3-Isobutoxy-2-cyclohexen-1-one 99% (10g) (AC291470100)

Category: Iron to Isopent
Description: 3-Isobutoxy-2-cyclohexen-1-one
Brand: Cole-Parmer


2-Acetyl-1-naphthol 99% (25g) (AC195000250)

Category: Acetylf to Acetylz
Description: 2-Acetyl-1-naphthol
Brand: Cole-Parmer


L(-)-2-Amino-1-butanol 98% (100ml) (AC244031000)

Category: Ametho to Aminob
Description: L(-)-2-Amino-1-butanol
Brand: Cole-Parmer


Oakton Mini-InfraPro Infrared Thermometer 8:1 (Model MT4) (MT4)

Category: Professional Compact Infrared Thermometers
Description: Oakton Mini-InfraPro Infrared Thermometers
Brand: Oakton


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought

Oakton 2-in-1 Waterproof Infrared Thermometer (7199OK)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Oakton 12.46 pH buffer, 500-mL  |  Next: Oakton 3000 ppm salt singles, NaCl calibration packets