Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-35634-30
Oakton Waterproof pHTestr 30 Pocket pH Tester
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-35634-30
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17101-45
NIST Traceable Calibration; pH Tester and Pens
$121.00 AUD/EACH/EACH
Usually ships in 23 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Oakton Waterproof pH Testr Pocket pH Testers
Share
|
Oakton Waterproof pHTestr 30 Pocket pH Tester
YO-35634-30
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Versatile, compact design works in a field or laboratory setting
±0.01 pH accuracy along with dual pH and selectable °C or °F temperature display
A complete meter with key features at an economical price
Replaceable, double-junction electrode sensors save time and money
IP67-rated housing sits flat on the table and floats in water
Waterproof pHTestr 30 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox
®
sensor housing resists most chemicals.
The pHTestr 30 has a replaceable sensor for easy maintenance. Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. Electrode sensor offers a clog-resistant Kynar
®
junction and a recessed, glass pH bulb to protect against breakage.
The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include a automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
What's Included:
Meter includes: protective plastic storage case, lanyard, and batteries.
Meter kit adds: two single-use pouches of pH 4.01, pH 7.00, pH 10.0 buffer, and rinse solution, one sample jar, and sturdy carrying case.
Specifications
Meter style
Tester
Dimensions (" L)
6 1/2
Dimensions (" Dia)
1 1/2
Range (pH)
-1 to 15 pH
Range (Temperature °C)
0 to 50
Range (Temperature °F)
32 to 122
Resolution (pH)
0.01
Accuracy (pH)
+/- 0.01
Temperature compensation
Automatic
Display
LCD
Calibration
Up to 3
Waterproof
Yes
Buffer recognition
USA, NIST
Power
Four 1.5V batteries
Battery life
500 hours
CE Compliance
Yes
Accessories
Qty
/
Price
Availability
Oakton buffer pack, 500-ml each of 4.01, 7.00, and 10.00 buffer
YO-05942-10
PACKAGE of 3
Call for availability.
Add to Cart
Batteries, 1.5V button cells, 6/PK
YO-09377-16
PACKAGE of 6
Call for availability.
Add to Cart
Replacement double-junction sensor for Oakton waterproof pH Testrs
3, 3+, 10, 20, 30
YO-35624-38
EACH
Call for availability.
Add to Cart
Soft carrying case for waterproof testers
YO-35624-45
EACH
Call for availability.
Add to Cart
Deluxe calibration kit
YO-35624-70
EACH
Call for availability.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Calibration; pH Tester and Pens
YO-17101-45
EACH
Usually ships in 23 days.
Add to Cart
Technical Resources
Instruction Manual
Oakton Catalogo en Espanol.pdf (8.3 MB)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Le secret pour assurer le bon fonctionnement d'un pH-mètre? Les électrodes!
The Secret to Well-Functioning pH Meters? (It's the Electrodes!)
Oakton pHTestr Sensor Module Replacement
Show All...
Double Junction pH Tester-The Next Generation of Long Life Pocket-Sized pH Meters
pH FAQs
Testing the pH Value of Pure Water
Testing the pH value of Meat
Testing the pH in Yogurt Production
Testing the pH value of Wine
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
pH Electrode Theory
Oakton® pH meter becomes a TV star!
Measuring the pH of Pure Water
pH Systems
pH Temperature Compensation
Temperature Compensation for pH Instruments
pH Measurement FAQs
Show Less...
Related Searches for Oakton Waterproof pHTestr 30 Pocket pH Tester
Waterproof Phtestr 30
Oakton Waterproof Phtestr 20
Hach Waterproof Non-Glass Isfet Ph Testers
Seattle Wa Ph Tester Kit
Oakton Ph Tester Phtestr 10
Ph Tester 30 Oakton
Oakton Ph Tester 35634 30 Modesto
Oakton Phtestr 30 Ph Meter Canning
Oakton Ph Tester Basic
Ph Tester 30
Related Products for Oakton Waterproof pHTestr 30 Pocket pH Tester
Image
Product Detail
SKU
Oakton Waterproof pHTestr 30 Pocket pH Tester and calibration kit
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pHTestr 30 Pocket pH Tester
Brand:
Oakton
Model Number:
pHTestr 30
YO-35634-90
More Details
Oakton Waterproof pHTestr 10 Pocket pH Tester
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pHTestr 10 Pocket pH Tester
Brand:
Oakton
Model Number:
pHTestr 10
YO-35634-10
More Details
Oakton Waterproof pHTestr 10 Pocket pH Tester and calibration kit
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pHTestr 10 Pocket pH Tester
Brand:
Oakton
Model Number:
pHTestr 10
YO-35634-70
More Details
Oakton Waterproof pHTestr 20 Pocket pH Tester and calibration kit
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pHTestr 20 Pocket pH Tester
Brand:
Oakton
Model Number:
pHTestr 20
YO-35634-80
More Details
Oakton Waterproof pHTestr 20 (-1.00 to 15.00 pH Range)
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pHTestr 20 Pocket pH Tester
Brand:
Oakton
Model Number:
pHTestr 20
YO-35634-20
More Details
Oakton Waterproof ORPTestr 10 Pocket ORP Tester
Category: pH/Ion Pocket Testers
Description: Oakton Waterproof ORPTestr 10 Pocket ORP Tester
Brand:
Oakton
Model Number:
ORPTestr
10
YO-35650-10
More Details
Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pHTestr 10 BNC Pocket pH Tester
Brand:
Oakton
Model Number:
pHTestr
10 BNC
YO-35634-14
More Details
Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
Category: Oakton EcoTestr Pocket Testers
Description: Oakton Waterproof EcoTestr pH 2 Pocket pH Tester
Brand:
Oakton
Model Number:
EcoTestr pH 2
YO-35423-10
More Details
Oakton Waterproof pH Spear Pocket pH Tester
Category: Oakton Waterproof pH Testr Pocket pH Testers
Description: Oakton Waterproof pH Spear Pocket pH Tester
Brand:
Oakton
Model Number:
pH Spear
YO-35634-40
More Details
Extech Waterproof PH100 Pocket pH Tester
Category: pH/Ion Pocket Testers
Description: Extech Waterproof PH100 Pocket pH Tester
Brand:
Extech
Model Number:
PH100
YO-53026-54
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Oakton Waterproof pHTestr 30 Pocket pH Tester
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Oakton Waterproof pHTestr 20 Pocket pH Tester and calibration kit
|
Next: Oakton Waterproof pHTestr 30 Pocket pH Tester and calibration kit
31125329-p-content_2 31125329pcontent2