Botulinum toxin detection kit (YO-36108-14)

YO-36108-14
$669.00 AUD/BOX

Usually ships in 14 days or sooner.
Qty

  • Quickly determine biological threat credibility
  • Results in 3- 30 minutes
  • Easy to read – two lines for positive; one line for negative
  • High Sensitivity and specificity
Accurately detect small, yet high threat amounts of biowarfare agents-without extensive sample preparation or additional expensive equipment. Test powder or liquid samples; kits are simple to use, results are quick and easy to read. Biohazard disposal bags and chain of custody labels are included for proper control and disposal or transport of the sample and test. Order simulation devices below for training purposes.
What’s included: 5" x 7" pouch includes all material to prepare and test a sample as well as package it for disposal or transport after testing.
Product Type Bacterial Test Kit
Tests for Botulinum Toxin





Technical Resources
 
Related Products for Botulinum toxin detection kit (YO-36108-14)
Image
  Product Detail
SKU

Ricin toxin detection kit (YO-36108-16)

Category: Bacteria test kits
Description: Biowarfare Agent Detection Devices (BADD)
Brand: Cole-Parmer


Anthrax detection kit (YO-36108-12)

Category: Bacteria test kits
Description: Biowarfare Agent Detection Devices (BADD)
Brand: Cole-Parmer


Replacement simulation detection devices (YO-36108-18)

Category: Bacteria test kits
Description: Biowarfare Agent Detection Devices (BADD)
Brand: Cole-Parmer


Assortment of detection devices mix of Anthrax, Botulinum, and Ricin (YO-36108-20)

Category: Bacteria test kits
Description: Biowarfare Agent Detection Devices (BADD)
Brand: Cole-Parmer


Test for indole reaction (YO-36151-14)

Category: Bacteria test kits
Description: Rapid Bacterial Test Kits
Brand: Cole-Parmer


Dissecting kit (YO-10915-00)

Category: Dissecting Instruments and Kits
Description: Dissecting Kit
Brand: Holiday Gift Ideas


Hach o-Phosphate Color Disc Test Kit (YO-05545-53)

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Hach o-Phosphate Color Disc Test Kit (YO-05545-57)

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Hach Chlorine, Free Color Disc Test Kit (YO-05545-68)

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Hach Chlorine, Total Color Disc Test Kit (YO-05545-70)

Category: Water Quality Test Kits
Description: Hach Color Disc Test Kits
Brand: HACH


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers

Do you have questions about this product?

get answers from real customers and in-house experts with AnswerBox.

2 Questions | 2 Answers
Displaying questions 1-2
  • Q:

    What are the values of sensitivity?

    Against that toxins is specificic?
    Asked on 2/20/2013 by Jose María from Valladolid, Spain

    1 answer

    • STAFF EXPERT

      A:

      The limit of detection for the two Botulinum toxins (A & B) are 33 ng/mL & 500 ng/mL. This test is not sensitive to the other toxin types (C-G).

      Answered on 2/22/2013 by Christina from Technical Support
  • Q:

    How many tests are there in each box of BADD.
    Asked on 12/28/2012 by Neeraja

    1 answer

    • STAFF EXPERT

      A:

      There are 10 tests per box.

      Answered on 12/28/2012 by Jen from Technical Support
Displaying questions 1-2

Do you have a question about this product? 

Product Reviews

Botulinum toxin detection kit (YO-36108-14)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Bottletop dispenser, no cover, 10-109 mL (844023)  |  Next: Bouffant Dispenser