Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Climatic Simulation Chamber 3 6 cu ft 200 240V
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-37756-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Environmental Chambers
Share
|
Climatic Simulation Chamber, 3.6 cu ft., 200-240V
YO-37756-20
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Meet International Conference on Harmonization (ICH) standards
DCT® cooling system prevents chamber icing and condensation
Preheating technology eliminates hot and cold spots
Symmetrical airflow system guarantees optimal temperature distribution
Four-channel, microprocessor-based controller ensures high levels of accuracy
All chambers feature an inner glass door for sample viewing without disrupting test conditions. Automatic defrost device protects the chamber during extended testing cycles. Access port 11⁄8" (29 mm) provides accessibility for chamber condition measurement or calibration. Included RS-422 interface transfers testing data to a PC for analysis. Temperature safety device features a visual alarm and complies with Class 3.1, DIN 12880 requirements.
Climatic Simulation Chambers
are ideal for pharmaceutical stability testing, QA/QC testing labs, and food packaging labs.
What's Included:
two stainless steel shelves, water connection kit (back-flow valve and inlet/outlet hoses), refrigerant, and power cord with plug.
Specifications
Product Type
Environmental Chambers
Capacity (cu ft)
3.6
Display
LCD
Temp accuracy
±0.2°F (±1°C)
Humidity range
10 to 80% RH
Humidity accuracy
±2% RH
Dimensions (chamber)
23 5/8" W x 19" H x 13 13/16" D
Dimensions (overall)
34 13/16" W x 41 5/16" H x 25 5/8" D
Chamber material
Stainless steel
Output
RS-422
Temp range
32 to 158°F (0 to 70°C)
Shelf positions
5
Power
200-240 VAC, 50/60 Hz
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Climatic Simulation Chamber, 3.6 cu ft., 200-240V
Climatic Test Chamber
Cu Ft Microwave Oven
24 Cu Ft Refrigerator
30 Cu Ft Refrigerators
Refrigerator 17 Cu Ft
6 Cu Ft Refrigerator
25 Cu Ft
40 Cu Ft Co2 Incubator
Bod Incubator 50 Cu Ft
6 Cu Ft Fridge
Related Products for Climatic Simulation Chamber, 3.6 cu ft., 200-240V
Image
Product Detail
SKU
Climatic Simulation Chamber, 24.7 cu ft., 200-240V
Category: Environmental Chambers
Description: Binder Climatic Simulation and Photostability Test Chambers
Brand:
Binder
Model Number:
9020-0108
YO-37756-21
More Details
Climatic/Photo Test Chamber 8.7 cu. ft., 200-240V
Category: Environmental Chambers
Description: Binder Climatic Simulation and Photostability Test Chambers
Brand:
Binder
Model Number:
9020-0160
YO-37756-25
More Details
Climatic/Photostability Test Chamber 24.7 cu. ft., 200-240V
Category: Environmental Chambers
Description: Binder Climatic Simulation and Photostability Test Chambers
Brand:
Binder
Model Number:
9020-0143
YO-37756-26
More Details
Laboratory space-saver refrigerator (single door); 11.5 CU FT; 200/230 (REL-1204D)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Laboratory Space-Saver Refrigerator with Single Door
Brand:
Thermo Scientific Revco
Model Number:
REL-1204D
YO-44185-15
More Details
Laboratory upright refrigerator (single door); 23.3 CU FT; 200/230V (REL-2304D)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Laboratory Upright Refrigerator with Single Door
Brand:
Thermo Scientific Revco
Model Number:
REL-2304D
YO-44185-25
More Details
ADA Compliant General Purpose Undercounter Refrigerator, 6 cu ft (6CAD)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6CAD
YO-44765-10
More Details
General Purpose Undercounter Refrigerator/Freezer, 6 cu ft (6CRF)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6CRF
YO-44765-45
More Details
Flammable Material Undercounter Refrigerator, 6 cu ft (6FAR)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6FAR
YO-44765-55
More Details
Flammable Material Undercounter Refrigerator/Freezer, 6 cu ft (6FRF)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6FRF
YO-44765-65
More Details
Hazardous Material Undercounter Refrigerator, 6 cu ft (6EAR)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6EAR
YO-44765-70
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Climatic Simulation Chamber, 3.6 cu ft., 200-240V
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Climatic Simulation Chamber, 24.7 cu ft., 200-240V
|
Next: Climatic/Photo Test Chamber 8.7 cu. ft., 200-240V
31125329-p-content_2 31125329pcontent2