Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
MAPA Temp Tec Heat Insulated Neoprene Gloves Large
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
YO-40211-31
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature-Resistant Gloves
Share
|
MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Large
YO-40211-31
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Two-tone wear indicator warns you when to replace gloves
These gloves are made of synthetic neoprene rubber for good chemical resistance. Easily maintain your grip with the nonslip, roughened surface. Withstand temperatures from -100 to 480°F (-73 to 249°C). The 14"L gloves feature integral cotton fleece liner.
Specifications
Product Type
Gloves
Material
Neoprene
Size
Large
Length
14 in
Color
Black and Blue
Thickness (mil)
Heavy gauge
Lining
Cotton jersey
Brand
Temp-Tec
Manufacturer number
332429
Model
332429
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
If the Glove Fits: The Right Safety Glove for your Application
Safety Gloves Sizing Guide
Safety Gloves Selection Guide
Show All...
Chemical Compatibility Chart for Safety Apparel
Safety Apparel Material Selection Guide
Cleanroom Garments
Show Less...
Related Searches for MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Large
Mapa Temp-Tec Heat-Insulated Neoprene Gloves
Mapa Temp Tec Gloves
Ansell Thermoprene Heat Resistant Neoprene Gloves
Mapa Temp-Tec Neoprene Gloves
Temp-Tec Temperature-Resistant Gloves
Large Neoprene Gloves
Restaurant Heat Resistant Rubber Gloves
Heat Resistant Cleanroom Gloves
Ansell Thermaprene Heat-Resistant
Small Neoprene Gloves
Related Products for MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Large
Image
Product Detail
SKU
MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Extra-large
Category: Temperature-Resistant Gloves
Description: MAPA Temp-Tec Heat-Insulated Neoprene Gloves
Brand:
Temp-Tec
Model Number:
332420
YO-40211-32
More Details
MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Medium
Category: Temperature-Resistant Gloves
Description: MAPA Temp-Tec Heat-Insulated Neoprene Gloves
Brand:
Temp-Tec
Model Number:
332428
YO-40211-30
More Details
MAPA Temp-Proof 333 Heat-Insulated Nitrile Gloves, size 9
Category: Temperature-Resistant Gloves
Description: MAPA Temp-Proof 333 Heat-Insulated Nitrile Gloves
Brand:
MAPA
Model Number:
333319
YO-40211-80
More Details
MAPA Knit, Double-Dipped, Stansoil® Neoprene Gloves; size large; 1 (334948)
Category: Reusable Neoprene Gloves
Description: MAPA
®
Knit, Double-Dipped, Stansoil
®
Neoprene Gloves
Brand:
Stansoil
Model Number:
334948
YO-40211-22
More Details
Ansell NeoTouch Neoprene Gloves, large, 100/box
Category: Disposable Gloves
Description: Ansell NeoTouch Neoprene Gloves
Brand:
Ansell
YO-86232-12
More Details
Ansell Thermaprene™ Heat-Resistant, Lined Neoprene Gloves, size 8,
Category: Temperature-Resistant Gloves
Description: Ansell Thermaprene Heat-Resistant, Lined Neoprene Gloves
Brand:
Thermaprene
Model Number:
19-026 214022
YO-81609-04
More Details
Ansell Thermaprene™ Heat-Resistant, Lined Neoprene Gloves, size 10,
Category: Temperature-Resistant Gloves
Description: Ansell Thermaprene Heat-Resistant, Lined Neoprene Gloves
Brand:
Thermaprene
Model Number:
19-026 214023
YO-81609-06
More Details
Ansell NeoTouch Neoprene Gloves, extra large, 100/box
Category: Disposable Gloves
Description: Ansell NeoTouch Neoprene Gloves
Brand:
Ansell
YO-86232-13
More Details
Ansell® Unlined Neoprene Gloves, size 9-9.5, 12 pair/pk
Category: Reusable Neoprene Gloves
Description: ANSELL Unsupported Neoprene Gloves
Brand:
Ansell
Model Number:
29-845 116241
YO-09112-24
More Details
Ansell® Unlined Neoprene Gloves, size 10-10.5, 12 pair/pk
Category: Reusable Neoprene Gloves
Description: ANSELL Unsupported Neoprene Gloves
Brand:
Ansell
Model Number:
29-845 116243
YO-09112-26
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Large
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Extra-large
|
Next: MAPA Temp-Tec Heat-Insulated Neoprene Gloves, Medium
31125329-p-content_2 31125329pcontent2