Pelican Unbreakable Instrument Case, 22" X 17" X 8" Interior

YO-40460-88
Loading price...

Call for availability.
Qty

  • Protect your valuable instrumentation—cases are absolutely air- and water-tight
  • Customize your case with prescored “Pick-N-Pluck” foam
  • Cases are made of unbreakable, corrosion-proof polypropylene
  • Built-in O-ring keeps case air- and water-tight
  • (Instruments are not included)
All Pelican cases are constructed of high-impact polypropylene and feature ABS plastic latches. A ¼" thick neoprene O-ring guarantees a water- and dust-proof seal. Each case includes a "Pick-N-Pluck” foam liner—customize your case by simply removing pieces of foam.
Models 40460-92 and -94 also include adjustable handle and rolling wheels for easy transport.
Interior dimensions 22"L x 17"W x 8"H
Product Type Instrument case
Exterior Dimensions 24-"L x 19-"W x 9"H
Model 1600-000-110
Brand Pelican
Manufacturer number 1600-000-110





Technical Resources
 
Related Products for Pelican Unbreakable Instrument Case, 22" X 17" X 8" Interior
Image
  Product Detail
SKU

Pelican Unbreakable Instrument case, 7-3/8" X 4-7/8" X 3-9/16" Interior

Category: Instrument Cases
Description: Pelican Unbreakable Instrument Cases
Brand: Pelican


Pelican Unbreakable Instrument case, 8-1/2" X 6" X 3-3/4" Interior

Category: Instrument Cases
Description: Pelican Unbreakable Instrument Cases
Brand: Pelican


Pelican Unbreakable Instrument Case, 17" X 11-3/4" X 6" Interior

Category: Instrument Cases
Description: Pelican Unbreakable Instrument Cases
Brand: Pelican


Pelican Unbreakable instrument case, Interior case 22-1/16"x17"x12-9/16"

Category: Instrument Cases
Description: Pelican Unbreakable Instrument Cases
Brand: Pelican


Pelican Unbreakable Instrument Case, Interior case 29"x17-7/8"x10-1/2" (1650-BLK)

Category: Instrument Cases
Description: Pelican Unbreakable Instrument Cases
Brand: Pelican


Pelican Unbreakable Instrument Case, 9-1/2" X 7-1/2" X 4-1/2" Interior

Category: Instrument Cases
Description: Pelican Unbreakable Instrument Cases
Brand: Pelican


Pelican Unbreakable Instrument Case, 12-1/4" X 9-1/4" X 5-1/4" Interior

Category: Instrument Cases
Description: Pelican Unbreakable Instrument Cases
Brand: Pelican


Pelican Black Case with foam and wheels model 1560, 20.37"x15.43"x9"

Category: Instrument Cases
Description: Pelican Watertight Instrument Cases
Brand: Pelican


Case, Black, 9.12 x 7.56 x 4.37in. O.D. with Foam; Inner Dimensions = 8.18" Long x 5.68" Wide x 3.62" Deep

Category: Instrument Cases
Description: Pelican Watertight Instrument Cases
Brand: Pelican


Case, Black, 13.37 x 11.62 x 6in. O.D. with Foam; Inner Dimensions = 11.81" Long x 8.87" Wide x 5.18" Deep

Category: Instrument Cases
Description: Pelican Watertight Instrument Cases
Brand: Pelican


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews