Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-41524-50
Standing Height Wall Assembly with Fume Hood - 13ft
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-41524-50
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Workstations
Share
|
Standing Height Wall Assembly with Fume Hood - 13ft (YO-41524-50)
YO-41524-50
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Complete systems make ordering easy!
These complete workstations include all of the components needed to install and get your laboratory up and running. Made of epoxy coated stainless steel, units are available in lengths from 3 to 16 feet, sure to fit your space requirements. Based on the most common configurations ordered, these systems are available in a variety of colors. Systems require assembly, wiring, and piping. All systems include 1” thick epoxy work surface and back splash. Wall assemblies also include a black epoxy backsplash.
Standing Height Wall Assemblies with Fume Hood
feature four cabinets complete with sink and constant volume/bypass fume hood. Fume hood is 48" wide and includes four remote control service fixtures and a cup sink. Fume hood requires blower and ductwork, order blowers separately. Black epoxy resin sink includes a deck-mount faucet, sink hangers, and plastic P-trap.
Specifications
Product Type
Cabinet assembly with fume hood
Dimensions (" W)
156
Dimensions (" H)
36-1/4
Dimensions (" D)
30
Shpg wt (lbs)
1825
Shpg wt (kg)
828
Description
13-ft four-cabinet assembly with fume hood
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related Searches for Standing Height Wall Assembly with Fume Hood - 13ft (YO-41524-50)
Cabinet Assembly with Fume Hood
Wall Mounted Chemical Fume Hood
Thermo Scientific Furniture & Fume Hoods
Chemical Fume Hood from Thermo Scientific
Cabinet Assembly with Fume
Alnor Furniture & Fume Hoods
Countertop Fume Hood
Explosion Proof Fume Hood
Benchtop Fume Hood with Drain
Laboratory Fume Hoods and Emergency Power
Related Products for Standing Height Wall Assembly with Fume Hood - 13ft (YO-41524-50)
Image
Product Detail
SKU
Standing Height Wall Assembly with Fume Hood - 14ft (YO-41524-55)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-55
More Details
Standing Height Wall Assembly with Fume Hood - 15ft (YO-41524-60)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-60
More Details
Standing Height Wall Assembly with Fume Hood - 16ft (YO-41524-65)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-65
More Details
Standing Height Wall Assembly with Wall Cabinets - 10ft (YO-41524-35)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-35
More Details
Standing Height Wall Assembly with Wall Cabinets - 11ft (YO-41524-40)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-40
More Details
Standing Height Wall Assembly with Wall Cabinets - 12ft (YO-41524-45)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-45
More Details
Sitting Height Complete Wall Assembly with countertop - 8ft, 6in (YO-41524-00)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-00
More Details
Sitting height complete wall assembly with countertop - 10ft, 6in (YO-41524-10)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-10
More Details
Basic electronic fume hood monitor (YO-33556-00)
Category: Airflow Monitors
Description: Fume Hood Airflow Monitors with Visual/Audible Alarm
Brand:
Alnor
YO-33556-00
More Details
Standing Height Wall Assmebly with Wall Cabinets - 9ft (YO-41524-30)
Category: Workstations
Description: Thermo Scientific Hamilton Laboratory Workbench Assemblies
Brand:
Thermo Scientific
YO-41524-30
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Standing Height Wall Assembly with Fume Hood - 13ft (YO-41524-50)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Standing height 14-ft wall assembly with cabinets, worksurface and sin (YO-41524-75)
|
Next: Standing Height Wall Assembly with Fume Hood - 14ft (YO-41524-55)
31125329-p-content_2 31125329pcontent2