Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Complete 6foot Fume Hood System With Acid And Solvent Storage Cabinet Epoxy Worksurface With Cup Sink
Click to enlarge
More Product Details
Specifications
Accessories
Build a complete system
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-41525-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Hamilton Constant Volume Bypass Fume Hoods
Share
|
Complete 6foot Fume Hood System With Acid And Solvent Storage Cabinet (YO-41525-20)
YO-41525-20
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Fume hood and cabinet assemblies make ordering easy
Systems come complete with hood, blower, work surface, and cabinets
These conveniently packaged systems take the guesswork out of ordering. Assemblies include a constant volume fume hood, a work surface, your choice of standard base or chemical storage cabinet, blower, vent kit, fillers, four service fixtures, molding, and corners. Included blowers are designed for use with 35 feet (10.7 m) of ductwork and one 90º angle; use existing ductwork or install new ductwork (not included). For information on these components, contact our Application Specialists.
Fume hoods and cabinets are made of durable powder-coated steel. Hoods and cabinets are available in 18 different colors; choose color when ordering from the chart below. The 4-ft (1.2-m) hood systems come with one cabinet; the 5-, 6-, and 8-ft (1.5-, 1.8-, and 2.4-m) hood systems come with two cabinets. All models must be wired and piped. Installation required (not included).
Specifications
Product Type
Ducted Fume Hood
Exterior Dimensions (" W)
72
Exterior Dimensions (" H)
96
Exterior Dimensions (" D)
31.25
Brand
Thermo Scientific Hamilton
Build a complete system
1
.
Hamilton Constant Volume Benchtop Fume Hood Fixtures
Details
2
.
Hamilton Constant Volume Benchtop Fume Hoods
Details
3
.
Hamilton Constant Volume Benchtop Fume Hoods Base Cabinets
Details
4
.
Hamilton Constant Volume Benchtop Fume Hoods Duct Transitions
Details
5
.
Hamilton Constant Volume Benchtop Fume Hoods Work Surfaces
Details
6
.
Hamilton Constant Volume Bypass Fume Hood Blowers
Details
Technical Resources
Instruction Manual.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Importance of Airflow Monitors for Fume Hoods
Selecting a chemical fume hood: it may be easier than you think!
To B2 or not to B2- a Biological Safety Cabinet Story
Show All...
Show Less...
Related Searches for Complete 6foot Fume Hood System With Acid And Solvent Storage Cabinet (YO-41525-20)
Chemical Fume Hood System
Acid Fume Hoods
Fisher Hamilton Concept Fume Hood
Fume Hood Dimensions
Thermo Scientific Fume Hoods
Hamilton Fume Hoods
Thermo Scientific Hamilton Fume Hood Assemblies
6 Fume Hood Fisher Scientific
Thermal Scientific Fume Hood
Thermo Scientific Hamilton Fume Hoods
Related Products for Complete 6foot Fume Hood System With Acid And Solvent Storage Cabinet (YO-41525-20)
Image
Product Detail
SKU
Complete 5foot Fume Hood System With Acid And Base Storage Cabinets Ep (YO-41525-10)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-10
More Details
Complete 8foot Fume Hood System With Acid And Base Storage Cabinet Epo (YO-41525-30)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-30
More Details
Complete 4foot Fume Hood System With Acid Storage Cabinet Epoxy Worksu (YO-41525-00)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-00
More Details
Complete 6foot Fume Hood System With 2 Base Storage Cabinet Epoxy Work (YO-41525-25)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-25
More Details
Complete 4foot Fume Hood System With General Base Storage Cabinet Epox (YO-41525-05)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-05
More Details
Complete 5foot Fume Hood System With 2 Base Storage Cabinet Epoxy Work (YO-41525-15)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-15
More Details
Complete 8foot Fume Hood System With 2 Base Storage Cabinet Epoxy Work (YO-41525-35)
Category: Hamilton Constant Volume Bypass Fume Hoods
Description: Hamilton Constant Volume Benchtop Fume Hoods Assemblies
Brand:
Thermo Scientific Hamilton
YO-41525-35
More Details
60" Standard Bypass Fume Hood with Standard Blower, 230 VAC/50 Hz (YO-09047-16)
Category: Ducted Fume Hoods
Description: Labconco
®
Protector
®
Premier
®
Fiberglass Fume Hoods
Brand:
Labconco
YO-09047-16
More Details
47" fiberglass fume hood with standard blower; 115 VAC/60 Hz (YO-33503-10)
Category: Ducted Fume Hoods
Description: Labconco 47- and 70-Inch Basic Fume Hoods
Brand:
Labconco
YO-33503-10
More Details
47" fiberglass fume hood with standard blower; 220 VAC/50 Hz (YO-33503-15)
Category: Ducted Fume Hoods
Description: Labconco 47- and 70-Inch Basic Fume Hoods
Brand:
Labconco
YO-33503-15
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Complete 6foot Fume Hood System With Acid And Solvent Storage Cabinet (YO-41525-20)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Complete 6foot Fume Hood System With 2 Base Storage Cabinet Epoxy Work (YO-41525-25)
|
Next: Complete 8foot Fume Hood System With 2 Base Storage Cabinet Epoxy Work (YO-41525-35)
31125329-p-content_2 31125329pcontent2