Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
14 1 2 ft seven cabinet island assembly with end sink and fixtures
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-41526-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Fume Hoods
Share
|
14-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-10)
YO-41526-10
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Increase workspace and storage with these complete assemblies
These complete island assemblies include all of the components needed to install and get your laboratory up and running. Made of epoxy coated stainless steel, units are available in lengths of 11-1/2 to 17-1/2 feet. These systems are available in a variety of colors, call our application specialists for color options. Systems require assembly, wiring, and piping. All systems include 1” thick epoxy work surface.
Island Assembly with End Sink
configuration is ideal for laboratories with overhead cabinets or limited overhead space. System includes seven cabinets and a black epoxy resin sink. Sink includes a deck-mount faucet, sink hangers, and plastic P-trap. Island also includes two double-faced electrical outlet boxes and four deck-mount gas hubs.
Specifications
Product Type
Island Assembly with sink
Dimensions (" W)
174
Dimensions (" H)
36-1/4
Dimensions (" D)
54
Shpg wt (lbs)
2104
Shpg wt (kg)
954
Description
141/2-ft seven-cabinet island assembly with end sink and fixtures
Brand
Thermo Scientific
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
To B2 or not to B2- a Biological Safety Cabinet Story
Importance of Airflow Monitors for Fume Hoods
Show Less...
Related Searches for 14-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-10)
Hamilton Lab Bench Island Assemblies
Island Assembly with Sink
Round Rubber Tubing 1 2
Cole-Parmer 1 2-Propanediol
Wika 4 1/2" Industrial Gauges
Eutech Instruments 1/2-Din Ph/Orp Control
1&1 2 Inch Rubber Tubing
Tygothane 1 2 Id
Peristaltic Pump 1 2 in Inlet
Surgical Rubber Tubing 1 2 Id
Related Products for 14-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-10)
Image
Product Detail
SKU
11-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-00)
Category: Fume Hoods
Description: Hamilton Lab Bench Island Assemblies
Brand:
Thermo Scientific
YO-41526-00
More Details
12-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-05)
Category: Fume Hoods
Description: Hamilton Lab Bench Island Assemblies
Brand:
Thermo Scientific
YO-41526-05
More Details
14-1/2 ft, nine cabinet island assembly with reagent shelf, sink, and (YO-41526-15)
Category: Fume Hoods
Description: Hamilton Lab Bench Island Assemblies
Brand:
Thermo Scientific
YO-41526-15
More Details
11-1/2 ft, seven cabinet island assembly with reagent shelf, sink, and (YO-41526-20)
Category: Fume Hoods
Description: Hamilton Lab Bench Island Assemblies
Brand:
Thermo Scientific
YO-41526-20
More Details
17-1/2 ft, eleven cabinet island assembly with reagent shelf, sink, an (YO-41526-25)
Category: Fume Hoods
Description: Hamilton Lab Bench Island Assemblies
Brand:
Thermo Scientific
YO-41526-25
More Details
2-ft Xpert Filtered Balance System With Guardian Airflow Monitor, 115V (YO-09102-10)
Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand:
Labconco
YO-09102-10
More Details
Thermo Scientific Nalgene fluorinated wide-mouth bottle, 1/2 gallon
Category: Wide-Mouth Bottles
Description: Thermo Scientific Nalgene Fluorinated 1/2-Gallon Wide-Mouth Bottle
Brand:
Thermo Scientific Nalgene
Model Number:
2124-0005
YO-62503-00
More Details
Eutech Instruments DO 2000 Controller, 1/2-DIN, 80 to 250 VAC
Category: 1/2-DIN Dissolved Oxygen and Bench Control
Description: Eutech Instruments DO 2000 1/2-DIN Dissolved Oxygen Controller
Brand:
Eutech Instruments
YO-53405-00
More Details
Eutech Instruments pH 2000 1/2-DIN Wall- Mount pH/ORP Controller
Category: Eutech Instruments 1/2-DIN pH/ORP Control
Description: Eutech Instruments pH 2000 Wall- and Panel-Mount pH/ORP Controllers/Transmitters
Brand:
Eutech Instruments
YO-56715-00
More Details
Front View Distillation Apparatus, Groups 0, 1, 2, and 3, right side, (K45000)
Category: Petroleum Testing
Description: Front View Distillation Apparatus, Groups 0, 1, 2, and 3
Brand:
Koehler
Model Number:
K45000
YO-59873-00
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
14-1/2 ft, seven cabinet island assembly with end sink and fixtures (YO-41526-10)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: 14-1/2 ft, nine cabinet island assembly with reagent shelf, sink, and (YO-41526-15)
|
Next: 14" Square SS Tray, w/handle (609086)
31125329-p-content_2 31125329pcontent2