Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
200 Ft
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-49005-52
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
200 Ft. (YO-49005-52)
YO-49005-52
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Related Searches for 200 Ft. (YO-49005-52)
40 Cu Ft Co2 Incubator
400 Od 200 Id Rubber Tubing
200 Tank
25 Cu Ft
Tube 52
Quick Disconnect Wolo Ba 52
Gilson Pipettes 20 200
Bod Incubator 50 Cu Ft
Epdm Peristaltic Pump Tubing Ft Myers
Rubber Tubing 1000 Ft Pric
Related Products for 200 Ft. (YO-49005-52)
Image
Product Detail
SKU
50 to 2500 mL sample volume range, 200 mm L x 25 mm OD, coarse disper (YO-04719-52)
Category: Homogenizer Probes, and Accessories
Description: IKA T-25 Homogenizer Rotor-Stator Generators
Brand:
IKA
YO-04719-52
More Details
High-purity PFA-450 tubing, 1/8"ID x 1/4"OD, 25 ft/pack (YO-06368-52)
Category: Rigid Tubing
Description: Cole-Parmer High-Purity PFA-450 Tubing
Brand:
Cole-Parmer
YO-06368-52
More Details
PTFE Tubing, 1.48 x 3mm, 25 ft/pk (YO-06605-52)
Category: PTFE Tubing
Description: Metric PTFE tubing
Brand:
Cole-Parmer
YO-06605-52
More Details
PTFE heat-shrink tubing, 0.560"ID, 12 ft/pack (YO-06851-52)
Category: Heat-Shrink Tubing
Description: PTFE tubing
Brand:
Cole-Parmer
YO-06851-52
More Details
5-ft XStream Hood, 115V, with 2 fixtures (YO-09099-52)
Category: Ducted Fume Hoods
Description: Labconco
®
Protector
®
XStream
™
Laboratory Hoods
Brand:
Labconco
YO-09099-52
More Details
4 ft XVS Ventilation Station, 31-3/4" working height (YO-09100-52)
Category: Laminar Flow Enclosures
Description: Labconco XVS Ventilation Stations
Brand:
Labconco
YO-09100-52
More Details
Organic Carbon Filter For 2-ft Xpert Systems (YO-09109-52)
Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand:
Labconco
YO-09109-52
More Details
Transducer F/ 32615-xx Series, 10 Ft, For 3" To 12" PVC, transducer 10 (YO-32615-52)
Category: Ultrasonic Flowmeters
Description: Cole-Parmer Ultrasonic Hybrid Doppler/Transit Time Flowmeters
Brand:
Cole-Parmer
YO-32615-52
More Details
Class II A2 5-ft Biosafety Cabinet with 8" sash opening and UV light, (YO-33515-52)
Category: Biosafety Cabinets (Class II)
Description: Labconco Purifier Logic Class II Biosafety Cabinets
Brand:
Labconco
YO-33515-52
More Details
Replacement DO probe with 10-ft cable for Oakton DO 6 meter (YO-35642-52)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Oakton
YO-35642-52
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
200 Ft. (YO-49005-52)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: 20" dual-gradient density polypropylene cartridge, 5 µm (155358-43)
|
Next: 200 g, Class 1 Calibration mass with certificate of calibration (82598CR)
31125329-p-content_2 31125329pcontent2