Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Hex Key For Use with Cole Parmer s Syringe Pump Infusion and Withdrawal design Touchscreen Control
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-74903-85
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
ColeParmer ® Touch Screen Syringe Pumps
Share
|
Sorry, this item is now discontinued but has a direct replacement.
You can compare specifications to the direct replacement YO-74905-85
,
browse similar items
or
Contact us for more information.
Hex Key For Use with Cole-Parmer's Syringe Pump, Infusion and Withdraw (788315) (YO-74903-85)
YO-74903-85
Loading price...
Discontinued
Qty
:
Save to shopping list
Technical Resources
Related technical articles (Roll over for summary)
The Ins and Outs of Metering Pumps
Metering Pump Selection Guide
How to Read a Pump Curve
Show All...
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Related Searches for Hex Key For Use with Cole-Parmer's Syringe Pump, Infusion and Withdraw (788315) (YO-74903-85)
Cole-Parmer Syringe Pumps
Cole-Parmer Syringe Accessories
Cole-Parmer Syringes
Cole-Parmer Syringe Guides
Cole-Parmer Hand Pumps
Cole-Parmer Drum Pumps
Cole-Parmer Diaphragm Pumps
Cole-Parmer Turbo Pumps without Integral Controller
Cole-Parmer Roller Pumps
Cole-Parmer Drum Pump Accessories
Related Products for Hex Key For Use with Cole-Parmer's Syringe Pump, Infusion and Withdraw (788315) (YO-74903-85)
Image
Product Detail
SKU
Microliter Rack for Cole-Parmer Syringe Pump, Infusion and Withdrawal (788302) (YO-74903-62)
Category: ColeParmer
®
Touch Screen Syringe Pumps
Description: Cole-Parmer
®
Touch Screen Syringe Pump Accessories
Brand:
Cole-Parmer
Model Number:
788302
YO-74903-62
More Details
Cole-Parmer dual-syringe infusion pump, 115 VAC
Category: Syringe Pumps
Description: Cole-Parmer Multi-Syringe Pumps
Brand:
Cole-Parmer
YO-74900-10
More Details
Cole-Parmer dual-syringe infusion pump, 230 VAC
Category: Syringe Pumps
Description: Cole-Parmer Multi-Syringe Pumps
Brand:
Cole-Parmer
YO-74900-15
More Details
Cole-Parmer dual-syringe infusion/withdrawal pump, 115 VAC
Category: Syringe Pumps
Description: Cole-Parmer Multi-Syringe Pumps
Brand:
Cole-Parmer
YO-74900-20
More Details
Cole-Parmer dual-syringe infusion/withdrawal pump, 230 VAC
Category: Syringe Pumps
Description: Cole-Parmer Multi-Syringe Pumps
Brand:
Cole-Parmer
YO-74900-25
More Details
Cole-Parmer ten-syringe infusion pump, 115 VAC
Category: Syringe Pumps
Description: Cole-Parmer Multi-Syringe Pumps
Brand:
Cole-Parmer
YO-74900-30
More Details
Cole-Parmer ten-syringe infusion pump, 230 VAC
Category: Syringe Pumps
Description: Cole-Parmer Multi-Syringe Pumps
Brand:
Cole-Parmer
YO-74900-35
More Details
Cole-Parmer ten-syringe infusion/withdrawal pump, 115 VAC
Category: Syringe Pumps
Description: Cole-Parmer Multi-Syringe Pumps
Brand:
Cole-Parmer
YO-74900-40
More Details
Cole-Parmer ten-syringe infusion/withdrawal pump, 230 VAC
Category: Syringe Pumps
Description: Cole-Parmer Multi-Syringe Pumps
Brand:
Cole-Parmer
YO-74900-45
More Details
Hex Key For Use with Cole-Parmer's Syringe Pump, Infusion and Withdraw
Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand:
Cole-Parmer
Model Number:
788315
YO-74905-85
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Hex Key For Use with Cole-Parmer's Syringe Pump, Infusion and Withdraw (788315) (YO-74903-85)
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Hex Key For Use with Cole-Parmer's Syringe Pump, Infusion and Withdraw
|
Next: HEXAAMINECOBALT III CHLORI 10G (YO-88205-93)
31125329-p-content_2 31125329pcontent2