Foam Insert for Accessories for Capture Fuming Station

YO-28616-24
Loading price...

Call for availability.
Qty

  • Quickly fixes latent fingerprints at the crime scene
  • Unique collapsible design is easy to transport and set up
  • Uses microburst or traditional CA fuming methods
  • Protects the user from hazardous fumes without ducting
Bring this portable fuming system directly to the crime scene, saving valuable laboratory time and space. System combines all the components needed for fingerprint fuming in one compact, easy-to-transport unit. A microprocessor-controlled heater heats the CA adhesive (Super Glue). The heater may be set in one degree increments between 120 to 350°F (49 to 77°C). An LCD displays the current or set-point temperature. A shatter-resistant, clear polycarbonate protective chamber, 8” W x 16” H x 8” D (20 x 41 x 20 cm), contains the fume as they react with the latent prints. A built-in exhaust blower pulls air from the chamber, through the filter and exhausts it into the room. A particulate/carbon exhaust filter traps smoke and CA fumes exhausted from the chamber.


Constructed of durable epoxy-coated steel with a stainless steel manifold to support evidence inside the chamber. A stainless steel hook, shelf and perforated tray are included. The CApture system runs on 115 VAC, 60 Hz, , 230 VAC, 50 Hz, or can be run on car battery when connected to a user-supplied inverter.
What's Included:
housing, chamber, hook, shelf, tray, built-in blower, particulate/carbon filter, 3-way heater/off/vent switch, leveling foot with locking pin, ten aluminum heater pans, 6-ft (1.8-m) cord and plug.

Product Type Forensic Fume Hood Accessories
Interior dimensions (" W) 8
Interior dimensions (" H) 16
Interior dimensions (" D) 8
Power (VAC) 115
Power (Hz) 60
Brand Labconco
Manufacturer number 3425500





Technical Resources
 
Related Products for Foam Insert for Accessories for Capture Fuming Station
Image
  Product Detail
SKU

Replacement Fuming Chamber for Capture Fuming Station

Category: Forensics
Description: Labconco CApture Portable Fuming System
Brand: Labconco


Replacment Filter Cartridge for Capture Fuming Station

Category: Forensics
Description: Labconco CApture Portable Fuming System
Brand: Labconco


Exhaust Connection Kit for Capture Fuming Station

Category: Forensics
Description: Labconco CApture Portable Fuming System
Brand: Labconco


Stainless Steel Fuming Shelf for Capture Fuming Station

Category: Forensics
Description: Labconco CApture Portable Fuming System
Brand: Labconco


Stainless Steel Fuming Hook for Capture Fuming Station

Category: Forensics
Description: Labconco CApture Portable Fuming System
Brand: Labconco


Perforated Shelf Tray for Capture Fuming Station

Category: Forensics
Description: Labconco CApture Portable Fuming System
Brand: Labconco


Labconco CApture Portable Fuming Station, 115 V

Category: Forensics
Description: Labconco CApture Portable Fuming System
Brand: Labconco


Labconco CApture Portable Fuming Station, 230 V

Category: Forensics
Description: Labconco CApture Portable Fuming System
Brand: Labconco


Carrying case with foam insert

Category: Air Sampling
Description: Air Sampling and Gas Analysis Accessories
Brand: Lab Products


Standard air blower, 1/6 hp, 115 VAC/60 Hz (YO-09078-00)

Category: Fume Hood Accessories
Description: Labconco Fume Hood Blowers
Brand: Labconco


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews