Laboratory Gas Regulator, Single Stage 590 CGA (Y11 C444B590)
YO-98202-66
Loading price...
Call for availability.
Qty
  • Designed for laboratory research using mildly corrosive gases
  • Diaphragm packless valve promotes system purity
  • Threadless seat design provides longer life
These stainless steel one- and two-stage high-purity regulators are designed for non-corrosive analytical, mildly corrossive and process applications requiring precise, stable delivery pressure control. A diaphragm packless outlet valve with a 1/4" compression tube fitting allows flow control and maintains system purity.
Single-stage regulators feature a convoluted stainless steel diaphragm seal that provides excellent regulating characteristics and allows for internal vacuum purging
Dual-stage regulators feature a a convoluted Hastelloy® C-22 diaphragm seal that prevents contamination by eliminating the need for a soft seal and provides a leak-rate design of less than 2 x 10–8 ccs helium. The two-stage models yield a delivery pressure change of less than 0.04/100 psi inlet change.
Product Type High Purity Gas Regulators
Max psi (inlet) 4000
Process connection 1/4" Swagelok®
Min operating temperature (º C) -40
Max operating temperature (º C) 74
Min operating temperature (º F) -40
Max operating temperature (º F) 165
Materials of construction 316 SS body & filter; SS gauges; Ni-plated brass bonnet; PCTFE® seat; Hastelloy C-22 diaphragm; SS outlet valve
Max Delivery Pressure (psig) 60
CGA fitting 590
Capacity (SCFH) 1250
Brand Cole-Parmer
Manufacturer number Y11 C444B590
Technical Resources
Related Products for Laboratory Gas Regulator, Single Stage 590 CGA (Y11 C444B590)
Image
  Product Detail
SKU
Laboratory Gas Regulator, Single Stage 330 CGA (Y11 C444B330)
Category: Pressure Regulators for Gases
Description: Cole-Parmer High-Purity Gas Regulators for Research
Brand: Cole-Parmer
Laboratory Gas Regulator, Single Stage 350 CGA (Y11 C444B350)
Category: Pressure Regulators for Gases
Description: Cole-Parmer High-Purity Gas Regulators for Research
Brand: Cole-Parmer
Laboratory Gas Regulator, Single Stage 580 CGA (Y11 C444B580)
Category: Pressure Regulators for Gases
Description: Cole-Parmer High-Purity Gas Regulators for Research
Brand: Cole-Parmer
Laboratory Gas Regulator, Single Stage 705 CGA (Y11 C444B705)
Category: Pressure Regulators for Gases
Description: Cole-Parmer High-Purity Gas Regulators for Research
Brand: Cole-Parmer
Laboratory Gas Regulator, Single Stage 330 CGA (Y11 C444D330)
Category: Pressure Regulators for Gases
Description: Cole-Parmer High-Purity Gas Regulators for Research
Brand: Cole-Parmer
Laboratory Gas Regulator, Dual Stage 590 CGA (Y12 C445B590)
Category: Pressure Regulators for Gases
Description: Cole-Parmer High-Purity Gas Regulators for Research
Brand: Cole-Parmer
Single-Stage Regulator, 180 Capacity, 590 CGA Fitting (Y11 244A590)
Category: Analytical Gas Regulators
Description: Single-Stage Regulators
Brand: Cole-Parmer
Single-Stage Regulator, 300 Capacity, 590 CGA Fitting (Y11 244B590)
Category: Analytical Gas Regulators
Description: Single-Stage Regulators
Brand: Cole-Parmer
Single-Stage Regulator, 480 Capacity, 590 CGA Fitting (Y11 244D590)
Category: Analytical Gas Regulators
Description: Single-Stage Regulators
Brand: Cole-Parmer
Laboratory Gas Regulator, Dual Stage 330 CGA (Y12 C445B330)
Category: Pressure Regulators for Gases
Description: Cole-Parmer High-Purity Gas Regulators for Research
Brand: Cole-Parmer
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews
Laboratory Gas Regulator, Single Stage 590 CGA (Y11 C444B590)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Previous: Laboratory Gas Regulator, Single Stage 580 CGA (Y11 C444B580)  |  Next: Laboratory Gas Regulator, Single Stage 705 CGA (Y11 C444B705)