WTW Multi 3420 Multiparameter Meter Only

YO-99575-17
Loading price...

Call for availability.
Qty

  • Dual-channel multiparameter meters—select exactly the features you need!
  • Graphical display of measured value and calibrated range for continuous measurement control
  • Sensor quality is displayed graphically for easy electrode diagnostics
  • Waterproof USB connection for ultra-flexible data output
WTW MultiLine meters are rugged, dual-channel instruments for pH, conductivity, and dissolved oxygen measurement. Multi 3420 display two channels using any combination of sensors. Order stand-alone meters or convenient kits for economical meter/sensor bundles.

MultiLine meters feature brilliant high resolution color displays. Measured readings, channel, and connector position information can be read at a glance. Unique display configuration also provides diagnostic information on sensor quality—easily tell when it is time for calibration or replacement! Plain display of calibrated measurement range ensures accurate readings.

Intelligent digital sensors (IDS) have proven advantages over other sensor types. Digital sensors are individually IDed and automatically log on to meters once connected. Sensors compare each following calibration with the initial calibration stored in the sensor providing valuable information on sensor health. Digital processing and data transfer of sensor measurement gives extremely precise measurements. Attach sensors via waterproof quick-lock sensor connections.

MultiLine meters also over state-of-the-art data management. Meters store up to 10,000 GLP-compliant measurements. Export data via waterproof USB or mini-USB interface. Waterproof connections are ideal for portable field measurements.

What's included: four AA batteries, CD-ROM, cable and power supply

Meter kits add: indicated pH, conductivity, or DO sensors, carrying case, calibration kit, stand and sample beaker.
Product Type Multiparameter Meter
Meter style Portable
Dimensions (" W) 7
Dimensions (" H) 3-1/8
Dimensions (" D) 2-1/4
Range (pH) 2.000 to 20.000
Range (mV) ±1250, ±2000
Range (Conductivity) 0 to 2000 mS
Range (TDS) 0 to 1999 mg/L
Range (Salinity) 0 to 70 ppt
Range (Resistivity) 0.00 to 20 M
Range (Dissolved oxygen) 0 to 20 mg/L; 0 to 200%
Range (Temperature °C) 5 to 105
Range (Temperature °F) 23 to 221
Resolution (pH) 0.1/0.01/0.001
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Resolution (Conductivity) 1.0 µS, 0.1, 1.0 mS
Resolution (TDS) 1 ppm; 0.01, 0.1 ppt
Resolution (Resistivity) 0.01 M ohm
Resolution (Dissolved Oxygen) 0.01 mg/L; 0.1% saturation
Resolution (Salinity) 0.1 ppt, 0.01%
Accuracy (pH) ±0.004 pH
Accuracy (Temperature) ±0.2°C
Accuracy (mV) ±0.2 mV or 0.05%, whichever is greater
Accuracy (Resistivity) ±0.5% ±1 digit
Accuracy (Conductivity) ±0.5% full-scale
Accuracy (TDS) ± 0.5% full-scale
Accuracy (Salinity) ±0.5% of reading
Accuracy (Dissolved Oxygen) ±0.5% of reading
Temperature compensation Automatic or manual
Display LCD
Interface USB
Calibration Up to 5 points
Waterproof Yes
Buffer recognition USA, NIST, DIN
Power Four AA batteries (included)
Datalogging (points) 10,000
CE Compliance Yes

Accessories
Qty/Price
Availability

EACH

Call for availability.

EACH

Call for availability.

EACH

Call for availability.
Calibrations
Qty/Price
Availability

EACH
Usually ships in 25 days.

EACH
Usually ships in 21 days.




Technical Resources
 
Related Products for WTW Multi 3420 Multiparameter Meter Only
Image
  Product Detail
SKU

WTW Multi 3420 Multiparameter Meter w/SenTix 940-3 and FDO 925-3 probes

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3420 Multiparameter Meters
Brand: WTW


WTW Multi 3420 Multiparameter Meter w/SenTix 940 and TetraCon 925 probes

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3420 Multiparameter Meters
Brand: WTW


WTW Multi 3420 Multiparameter Meter w/SenTix 940-3 and TetraCon 925-3 probes

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3420 Multiparameter Meters
Brand: WTW


WTW Multi 3420 Multiparameter Meter w/SenTix 980 and LR 925/01 probes

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3420 Multiparameter Meters
Brand: WTW


WTW Multi 3420 Multiparameter Meter; SenTix 940-3/TetraCon 925-3/FDO 925-3

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3420 Multiparameter Meters
Brand: WTW


WTW Multi 3420 Multiparameter Meter w/SenTix 950 and TetraCon 925 probes

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3420 Multiparameter Meters
Brand: WTW


WTW Multi 3410 Multiparameter Meter Only

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3410 Multiparameter Meters
Brand: WTW


WTW Multi 3410 Multiparameter Meter w/SenTix 940 pH probe

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3410 Multiparameter Meters
Brand: WTW


WTW Multi 3410 Multiparameter Meter w/SenTix 940-3 pH probe

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3410 Multiparameter Meters
Brand: WTW


WTW Multi 3410 Multiparameter Meter w/SensoLyt 900-6 pH probe

Category: Handheld Multiparameter Meters
Description: WTW MultiLine 3410 Multiparameter Meters
Brand: WTW


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews