Home  >  pH/Ion Meter Accessories >
  

Pipette filler, safety bulb

YO-99574-53
Loading price...

Call for availability.
Qty
Model 14651-00
Brand Hach





Technical Resources
 
Related Products for Pipette filler, safety bulb
Image
  Product Detail
SKU

Safety Pipette Filler bulb with removable Valve (FLIP-001)

Category: Pipette Fillers
Description: Safety Pipette Filler with Removable Valve
Brand: Cole-Parmer


Jencons Smoothie Pipette Filler, Vanilla (475-360)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Jencons Smoothie Pipette Filler, Strawberry (475-361)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Jencons Smoothie Pipette Filler, Banana (475-362)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Jencons Smoothie Pipette Filler, Lemon Lime (475-363)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Jencons Smoothie Pipette Filler, Blueberry (475-364)

Category: Pipette Fillers
Description: Jencons Smoothie Pipette Fillers
Brand: Jencons Smoothie


Safetypette pipette filler (475-165)

Category: Pipette Fillers
Description: Cole-Parmer Safetypette
Brand: Cole-Parmer


50.0 mL Silicone pipette filler (SILRED-001)

Category: Pipette Fillers
Description: Silicone Pipette Filler
Brand: Cole-Parmer


Cole-Parmer Pipette Controller, filler, fits standard pipettes; 50 mL capacity

Category: Pipette Fillers
Description: Cole-Parmer Pipette Filler
Brand: Cole-Parmer


Pipette Pumps, yellow, 0.2 mL (37896)

Category: Pipette Fillers
Description: Pipette Pumps
Brand: Scienceware


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews