Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
H B Instrument Soil Hydrometer ASTM 152H 5 to 60 Grams Plain Form
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-08297-42
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17107-00
NIST-Traceable Calibration; Hydrometer with test data, one-point (midpoint of Hydrometer's range)
$242.00 AUD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Hydrometers Special Applications
Share
|
H-B Instrument Soil Hydrometer, ASTM 152H, —5 to 60 Grams, Plai
YO-08297-42
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
NIST Traceable
Temperature of Standardization 68°F/68°F
Conform to ASTM Specifications
Metal Ballast
Individually Serialized
Ideal for determining the specific gravity of soil, meets ASTM specifications. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
Specifications
Product Type
Hydrometers, Soil
Manufacturer number
152H
Form
Plain
Model
152H
Brand
H-B Instrument Company
Accessories
Qty
/
Price
Availability
Hydrometer Case for 380 mm Length Hygrometer
YO-08285-97
EACH
Call for availability.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Hydrometer with test data, one-point (midpoint of Hydrometer's range)
YO-17107-00
EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Hydrometer Selection Guide
How to Read a Hydrometer
Brix Scale (sucrose)
Show All...
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show Less...
Related Searches for H-B Instrument Soil Hydrometer, ASTM 152H, —5 to 60 Grams, Plai
Soil Hydrometers
H-B Instruments Soil Analysis Hydrometers
H-B Instrument Company Hydrometers Special Applications
Hydrometer Astm
H-B Instrument Company Hydrometers
H-B Instrument Company Hydrometers for Salt
H-B Instrument Company Hydrometer Accessories
Soil Temperature Instrument
Instrumentation Companies
West and Company Weather Instruments
Related Products for H-B Instrument Soil Hydrometer, ASTM 152H, —5 to 60 Grams, Plai
Image
Product Detail
SKU
H-B Instrument Soil Hydrometer, ASTM 151H, 0.995 to 1.038 Specific Gra
Category: Hydrometers Special Applications
Description: H-B Instrument Soil Analysis Hydrometers
Brand:
H-B Instrument Company
Model Number:
151H
YO-08297-40
More Details
H-B Instrument Hydrometer, ASTM 82H, 0.650 to 0.700 Specific Gravity,
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
82H
YO-08297-81
More Details
H-B Instrument Hydrometer, ASTM 86H, 0.85 to 0.90 Specific Gravity, 0.
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
86H
YO-08297-89
More Details
H-B Instrument Hydrometer, ASTM 87H, 0.90 to 0.95 Specific Gravity, 0.
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
87H
YO-08297-91
More Details
H-B Instrument Hydrometer, ASTM 88H, 0.95 to 1.00 Specific Gravity, 0.
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
88H
YO-08297-93
More Details
H-B Instrument Hydrometer, ASTM 102H, 0.650 to 0.700 Specific Gravity,
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
102H
YO-08297-97
More Details
H-B Instrument Hydrometer, ASTM 104H, 0.75 to 0.80 Specific Gravity, 0
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
104H
YO-08298-01
More Details
H-B Instrument Hydrometer, ASTM 106H, 0.85 to 0.90 Specific Gravity, 0
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
106H
YO-08298-05
More Details
H-B Instrument Hydrometer, ASTM 107H, 0.90 to 0.95 Specific Gravity, 0
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
107H
YO-08298-07
More Details
H-B Instrument Hydrometer, ASTM 103H, 0.700 to 0.750 Specific Gravity,
Category: Hydrometers for Specific Gravity
Description: H-B Instrument Specific Gravity ASTM Hydrometers for Liquids Lighter than Water
Brand:
H-B Instrument Company
Model Number:
103H
YO-08297-99
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
H-B Instrument Soil Hydrometer, ASTM 152H, —5 to 60 Grams, Plai
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: H-B Instrument Soil Hydrometer, ASTM 151H, 0.995 to 1.038 Specific Gra
|
Next: H-B Instrument Specific Gravity Hydrometer Set, 8 Plain Form, 0.700 to
31125329-p-content_2 31125329pcontent2