Replacement Oxygen Sensor, 0 to 25%

YO-86515-19
Loading price...

  • Three alarms functions—audible, visual, and vibration—make alarm conditions clear
The monitor features a robust, rubber-coated weatherproof polymer housing rated to IP65 that will stand up to the typical abuse in an industrial environment. Easily operated using just two buttons. Its small size makes it comfortable and easy to wear all day on a belt, pocket, or lapel. A loud audible alarm, two bright visual alarms, and strong vibrating alarm alert you to hazardous concentrations of gas. Built-in event logger stores up to 60 alarm activations or other events.
Product Type Replacement Toxic Gas Sensors
Gas Oxygen
Manufacturer number 6810881
CE Compliance Yes
Brand Drager





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Related Products for Replacement Oxygen Sensor, 0 to 25%
    Image
      Product Detail
    SKU

    Polarization Dissolved Oxygen Sensor for BioBundle Fermentation System, 2-pin low-drift

    Category: BioBundle Accessories
    Description: Polarization Dissolved Oxygen Sensors
    Brand: BioBundle


    Dissolved Oxygen Sensor for BioBundle ™ Fermentation System, 110 mm

    Category: BioBundle Accessories
    Description: Dissolved Oxygen Sensors
    Brand: BioBundle


    Dissolved Oxygen Sensor for BioBundle ™ Fermentation System, 154 mm

    Category: BioBundle Accessories
    Description: Dissolved Oxygen Sensors
    Brand: BioBundle


    Dissolved Oxygen Sensor for BioBundle ™ Fermentation System, 235 mm

    Category: BioBundle Accessories
    Description: Dissolved Oxygen Sensors
    Brand: BioBundle


    Dissolved Oxygen Sensor for BioBundle ™ Fermentation System, 325 mm

    Category: BioBundle Accessories
    Description: Dissolved Oxygen Sensors
    Brand: BioBundle


    Dissolved Oxygen Sensor for BioBundle ™ Fermentation System, 590 mm

    Category: BioBundle Accessories
    Description: Dissolved Oxygen Sensors
    Brand: BioBundle


    Replacement oxygen sensor (YO-86419-36)

    Category: Air Sampling
    Description: Air Sampling and Gas Analysis Accessories
    Brand: Lab Products


    Replacement oxygen sensor (YO-10112-16)

    Category: Air Sampling
    Description: Air Sampling and Gas Analysis Accessories
    Brand: Lab Products


    Replacement oxygen sensor (24-0788)

    Category: Air Sampling
    Description: Air Sampling and Gas Analysis Accessories
    Brand: Lab Products


    Global Water Dissolved Oxygen Sensor (WQ401)

    Category: Dissolved Oxygen (DO) Probes and Sensors
    Description: Global Water DO Sensor
    Brand: Global Water


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    Replacement Oxygen Sensor, 0 to 25%
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Replacement oxygen sensor (YO-86419-36)  |  Next: Replacement pads (F36251)