Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-17400-70
Western States Laboratory Filtering Centrifuge
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-17400-70
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Centrifuges
Share
|
Western States Laboratory Filtering Centrifuge
YO-17400-70
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Rugged and versatile lab centrifuge is easy to use, no tools required!
Create accurate, repeatable results with electronic speed control and digitally run timer
Generate forces of up to 2130 gravities!
Recover 100% of the retained solids in your sample.
Sturdy stainless steel design ensures years of use and meets cGMP compliance
NEMA-4x rated for excellent corrosion resistance
Cole-Parmer offers almost every type of centrifuge available, including small benchtop, personal microcentrifuges, general purpose, even specialty centrifuges developed for crude oil tests or pharmaceutical filtration. Contact us if you cannot find exactly what you are looking for- we can help you find a solution.This compact, sanitary filtering centrifuge is designed for solid and liquid separation: ideal in the for washing and extracting food ingredients, crystals, botanical extracts, pharmaceuticals, and polysaccharides. Vaportight vessel features a hinged cover with safety interlock to prevent cover opening while unit is in use. View rotating sample through the sight glass at the top. Special fittings allow for complete liquid drainage and 20-watt halogen light ensures proper sanitation while unit is in use.
Removable basket is easy to clean and sterilize, and is highly perforated, eliminating the need for backing screens or tools-just carry to your workbench. Basket is machined from a solid block of 316 stainless steel- no cracks or crevices to contain microbes. Rugged design can handle even the most dense products- including powdered metals. Heavy duty bearings isolate the motor from the basket, and triple isolated mounts eliminate vibrations.
Specifications
Product Type
Centrifuge
Max rpm
5000
Speed range
0 to 5000 rpm
Max capacity
Solids holding volume = 56.5 in^3 (927cm^3)
Refrigerated
No
Wetted parts (Housing)
316 Stainless Steel
Power (VAC)
110
Rotor (included)
Yes
Timer
99 min, 59 sec or 99 hr, 59 min
Manufacturer number
072755
Brand
Western States
Dimensions
26" x 39" x 18"
Accessories
Qty
/
Price
Availability
Portable Stainless Steel Cart for Laboratory Filtering Centrifuge
YO-17400-72
EACH
Call for availability.
Add to Cart
Filter paper - 320 grade for laboratory filtering centrifuge (17400-70) 10/pk
YO-17400-74
PACKAGE
Call for availability.
Add to Cart
Filter paper - 950 grade for laboratory filtering centrifuge (17400-70)- 10/pk
YO-17400-76
PACKAGE
Call for availability.
Add to Cart
Filter paper - 1278 grade for laboratory filtering centrifuge (17400-70)- 10/pk
YO-17400-78
PACKAGE
Call for availability.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Isolating Genomic DNA from Whole Blood
Common Centrifuge Terms
Basics of Centrifugation
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Related Searches for Western States Laboratory Filtering Centrifuge
Western States Laboratory Equipment
Western States Centrifuge Troubleshooting Tach
Western States Filtering Centrifuges
Filtering Centrifuges
Micropore Filter Centrifuge Tube
Laboratory Filter Funnel
Laboratory Filter Paper
Laboratory Filter Capsules
Laboratory Filtering Flasks
Laboratory Filter Holders
Related Products for Western States Laboratory Filtering Centrifuge
Image
Product Detail
SKU
Portable Stainless Steel Cart for Laboratory Filtering Centrifuge
Category: Centrifuges
Description: Western States Laboratory Filtering Centrifuge
Brand:
Western States
YO-17400-72
More Details
Filter paper - 320 grade for laboratory filtering centrifuge (17400-70
Category: Centrifuges
Description: Western States Laboratory Filtering Centrifuge
Brand:
Western States
YO-17400-74
More Details
Filter paper - 950 grade for laboratory filtering centrifuge (17400-70
Category: Centrifuges
Description: Western States Laboratory Filtering Centrifuge
Brand:
Western States
YO-17400-76
More Details
Filter paper - 1278 grade for laboratory filtering centrifuge (17400-7
Category: Centrifuges
Description: Western States Laboratory Filtering Centrifuge
Brand:
Western States
YO-17400-78
More Details
9" Centrifuge Brush
Category: Laboratory Brushes
Description: Centrifuge and Micro Brushes
Brand:
Cole-Parmer
Model Number:
610215
YO-84551-90
More Details
9" Centrifuge Brush, 12/pk (610217)
Category: Laboratory Brushes
Description: Centrifuge and Micro Brushes
Brand:
Cole-Parmer
Model Number:
610217
YO-84551-91
More Details
9" Centrifuge Brush, 12/pk (610218)
Category: Laboratory Brushes
Description: Centrifuge and Micro Brushes
Brand:
Guaranteed In Stock Program
Model Number:
610218
YO-84551-92
More Details
12" Centrifuge Brush, 12/pk
Category: Laboratory Brushes
Description: Centrifuge and Micro Brushes
Brand:
Cole-Parmer
Model Number:
610222
YO-84551-93
More Details
6" Micro Brush, 12/pk
Category: Laboratory Brushes
Description: Centrifuge and Micro Brushes
Brand:
Cole-Parmer
Model Number:
710338
YO-84551-94
More Details
7" Micro Brush, 12/pk
Category: Laboratory Brushes
Description: Centrifuge and Micro Brushes
Brand:
Cole-Parmer
Model Number:
720340
YO-84551-95
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Western States Laboratory Filtering Centrifuge
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: West condenser, medium length, 190 mm, 14/20 joint (YO-34705-94)
|
Next: Wet/dry soil sampling tube 12" long (PN012)
31125329-p-content_2 31125329pcontent2