Home  >  Chemicals Sa to Sz >
  

Salicylic Acid (Crystalline or Powder/Certified ACS) (500g)

YO-88022-00
Loading price...

Usually ships in 14 days.
Qty

  • Assay—≥99.0%
  • Melting Point—158.0°-161.0°C
  • Residue on Ignition—≤0.010%
  • Chloride—0.001%
  • Substances Darkened by H2SO4To pass test
  • Sulfate—≤0.003%
  • Heavy Metals (as Pb)—≤5 ppm
  • Iron—≤2ppm

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Chemical Reagents
Purity grade Certified ACS
CAS Number 69-72-7
Form Crystals / Powder
Formula weight 138.12
Empirical formula C7H6O3
Linear formula 2-HOC6H4HCOOH
Synonyms 2-Hydroxybenzoic Acid
Packaging Amber Glass
Storage symbol Gray
Hazard symbol Harmful
Quantity 500g





Technical Resources
 
Related Products for Salicylic Acid (Crystalline or Powder/Certified ACS) (500g)
Image
  Product Detail
SKU

L-(+)-Tartaric Acid (Powder/Certified ACS) (500g)

Category: Ta to Terpine
Description: L(+)-Tartaric acid
Brand: Cole-Parmer


m-Phosphoric Acid, Glacial (Crystalline or Flakes/Certified ACS) (500g)

Category: Phenyls to Phospho
Description: m-Phosphoric Acid, Glacial
Brand: Cole-Parmer


Starch, Soluble (Powder/Certified ACS) (500g)

Category: Solketa to Sulfom
Description: Starch, Soluble
Brand: Cole-Parmer


Hydrazine Sulfate (Powder/Certified ACS) (500g)

Category: Homatro to Hydroq
Description: Hydrazine Sulfate
Brand: Cole-Parmer


Potassium Carbonate Sesquihydrate (Powder/Certified ACS) (500g)

Category: Potassium
Description: Potassium Carbonate Sesquihydrate
Brand: Cole-Parmer


Silver Sulfate (Powder/Certified ACS) (500g) (S190500)

Category: Shikimi to Soda
Description: Silver Sulfate
Brand: Cole-Parmer


Sodium Molybdate Dihydrate (Cryst. Powder/Certified ACS) (500g) (S336500)

Category: Sodiumh to Sodiumo
Description: Sodium Molybdate Dihydrate
Brand: Cole-Parmer


Thioacetamide (Cryst. Powder/Certified ACS) (500g) (T102500)

Category: Tetrar to Thymine
Description: Thioacetamide
Brand: Cole-Parmer


Zinc Oxide (Powder/Certified ACS) (500g) (Z52500)

Category: Za to Zz
Description: Zinc Oxide
Brand: Cole-Parmer


Salicylic Acid (Crystalline/USP) (2.5kg)

Category: Chemicals Sa to Sz
Description: Salicylic Acid
Brand: Cole-Parmer


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

Salicylic Acid (Crystalline or Powder/Certified ACS) (500g)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Salicylhydrazide 98% (25g)  |  Next: Salicylic Acid (Crystalline/USP) (2.5kg)