Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Viton Drum Seal Set
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-75650-56
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Hand-Operated Drum Pumps
Share
|
Viton Drum Seal Set
YO-75650-56
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Pump wide selection of fluids from 5 gallon pails to 55 gallon barrels
Choice of four seal materials to handle over 800 liquids and chemicals
Guaranteed for 1 year. Life expectancy 7–10 years
Internal safety relief valve prevents overpressurization
Drums are Resource Conservation and Recovery Act (RCRA) ready
The GoatThroat pumps are designed to handle a wide range of liquids—wetted parts are 100% nonreactive, food-grade polypropylene. Choose nitrile seals for petroleum-based products including oil, diesel fuel, and glycerin. The EPDM seals are ideal for general-purpose chemicals including some acids, acetates, alcohols, and phosphates. Santoprene
®
seals are ideal for acetic acid, acetone, and hydrocyanic acid among others. Viton
®
seals were developed for more aggressive chemicals including bromine, sulfuric acid, ethylene, nitric acid, gasoline, and pesticides.
What's Included:
medium drum seal (2.175" [5.5 cm]), four siphon tube segments, three siphon tube connectors, and one foot piece, 2" NPT fitting, rubber compression fitting, Molykote III, PTFE tape.
Specifications
Product Type
Drum Pump Seal Sets
Brand
GoatThroat
Technical Resources
Related technical articles (Roll over for summary)
Closed System Technology Drives the Trend Toward Safer, More Cost-Efficient Chemical Dispensing
How to Read a Pump Curve
Guide to Liquid Pumps
Show All...
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Related Searches for Viton Drum Seal Set
Drum Pump Seal Sets
Eppendorf Pipette Piston Seal Set
Santoprene Hand-Operated Drum Pumps
Goatthroat Hand-Operated Drum Pumps
Goatthroat Drum and Hand Pumps
Vacuum Sealing Systems
Cole-Parmer Complete Drum and Laboratory Pumps
Rotary Evaporator Vacuum Seal Brinkmann
Laboratory Pumps
Rotary Evaporator Replacement
Related Products for Viton Drum Seal Set
Image
Product Detail
SKU
Nitrile Drum Seal Set
Category: Hand-Operated Drum Pumps
Description: GoatThroat Spillproof Barrel Pumps and Accessories
Brand:
GoatThroat
Model Number:
DS100
YO-75650-50
More Details
Epdm Drum Seal Set
Category: Hand-Operated Drum Pumps
Description: GoatThroat Spillproof Barrel Pumps and Accessories
Brand:
GoatThroat
Model Number:
DS200
YO-75650-52
More Details
Santoprene Drum Seal Set
Category: Hand-Operated Drum Pumps
Description: GoatThroat Spillproof Barrel Pumps and Accessories
Brand:
GoatThroat
Model Number:
DS200S
YO-75650-54
More Details
Complete Laboratory Drum Pump Set, 27" in length, 18.5 GPM, Polypropyl
Category: Electric and Air Powered Drum/Tote Pumps
Description: Complete Drum and Laboratory Pumps
Brand:
Cole-Parmer
Model Number:
B2-PP-SL-25"
YO-70510-12
More Details
Complete Laboratory Drum Pump Set, 27" in length, 17 GPM, Stainless St
Category: Electric and Air Powered Drum/Tote Pumps
Description: Complete Drum and Laboratory Pumps
Brand:
Cole-Parmer
Model Number:
B2-PP-SL-25"
YO-70510-14
More Details
Complete Laboratory Drum Pump Set, 39" in length, 18.5 GPM, Polypropyl
Category: Electric and Air Powered Drum/Tote Pumps
Description: Complete Drum and Laboratory Pumps
Brand:
Cole-Parmer
Model Number:
B2-PP-SL-25"
YO-70510-17
More Details
Complete Laboratory Drum Pump Set, 39" in length, 17 GPM, Stainless St
Category: Electric and Air Powered Drum/Tote Pumps
Description: Complete Drum and Laboratory Pumps
Brand:
Cole-Parmer
Model Number:
B2-PP-SL-25"
YO-70510-19
More Details
PP Pressure Gauge, Viton diaphragm seal, 1/4" NPT(F); 0 to 15 psi
Category: Mechanical Pressure Gauges
Description: PVDF and PP Mechanical Pressure Gauges
Brand:
PTFE
Model Number:
KC701LDS25(15)
YO-68000-25
More Details
PP Pressure Gauge, Viton diaphragm seal, 1/4" NPT(F); 0 to 30 psi
Category: Mechanical Pressure Gauges
Description: PVDF and PP Mechanical Pressure Gauges
Brand:
PTFE
Model Number:
KC701LDS25(30)
YO-68000-27
More Details
PP Pressure Gauge, Viton diaphragm seal, 1/4" NPT(F); 0 to 60 psi
Category: Mechanical Pressure Gauges
Description: PVDF and PP Mechanical Pressure Gauges
Brand:
PTFE
Model Number:
KC701LDS25(60)
YO-68000-29
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Viton Drum Seal Set
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Vistek™ Balance Isolation Platform, 8"W x 10"D
|
Next: Viton Gasket, 10 MESH, Size 1-1/2" Tri-Clamp (YO-30536-06)
31125329-p-content_2 31125329pcontent2