Thermo Scientific Orion pH Electrode Storage Solution, 5 gallon (19L) cubitainer

YO-05730-66
Loading price...

Call for availability.
Qty
    Thermo Scientific Orion pH electrode storage solutions restore proper electrode performance and prolong the useful life of the electrode.
    Manufacturer number 9100CB





    Technical Resources
     
    Related Products for Thermo Scientific Orion pH Electrode Storage Solution, 5 gallon (19L) cubitainer
    Image
      Product Detail
    SKU

    Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottles

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Storage Solutions
    Brand: Thermo Scientific Orion
    Model Number: 910060


    Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Storage Solutions
    Brand: Thermo Scientific Orion
    Model Number: 910001


    Thermo Scientific Orion ROSS Storage Soluton, 475-mL bottle

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion ROSS Electrode Storage Solution
    Brand: Thermo Scientific Orion
    Model Number: 810001


    Thermo Scientific Orion pH Electrode Cleaning Solution Kit

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Cleaning Solutions
    Brand: Thermo Scientific Orion
    Model Number: 900020


    Thermo Scientific Orion pH electrode cleaning solution A, 4 x 30 mL bottles

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Cleaning Solutions
    Brand: Thermo Scientific Orion
    Model Number: 900021-WA


    Thermo Scientific Orion pH electrode cleaning solution B, 4 x 60 mL bottles

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Cleaning Solutions
    Brand: Thermo Scientific Orion
    Model Number: 900022-WA


    Thermo Scientific Orion pH electrode cleaning solution C, 4 x 30 mL bottles

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Cleaning Solutions
    Brand: Thermo Scientific Orion
    Model Number: 900023


    Thermo Scientific Orion pH electrode cleaning solution D, 4 x 60 mL bottles

    Category: pH/Ion Buffers, and Storage Solutions
    Description: Thermo Scientific Orion Electrode Cleaning Solutions
    Brand: Thermo Scientific Orion
    Model Number: 900024


    Thermo Scientific Orion pH Electode, Sealed/DJ/Epoxy; WP BNC

    Category: pH/Ion Laboratory Electrodes
    Description: Thermo Scientific Orion Standard Ag/AgCl pH Electrodes
    Brand: Thermo Scientific Orion
    Model Number: 9156DJWP


    Thermo Scientific Orion pH Probe, Refillable/SJ/Glass; WP BNC

    Category: pH/Ion Laboratory Electrodes
    Description: Thermo Scientific Orion Standard Ag/AgCl pH Electrodes
    Brand: Thermo Scientific Orion
    Model Number: 9102BNWP


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews

    Thermo Scientific Orion pH Electrode Storage Solution, 5 gallon (19L) cubitainer
    Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
    Popularity |  Favorites |  Title |  Price |  Part Number |  Category
    Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

    Previous: Thermo Scientific Orion pH Electrode Storage Solution, 1 pint (475-mL)  |  Next: Thermo Scientific Orion pH Electrode Storage Solution, 5 x 60-mL bottles