Thermo Scientific GENESYS ™ 10S Bio UV/Visible Spectrophotometer, 6 cell

YO-02654-00
Loading price...

Call for availability.
Qty

  • Versatile, accurate UV/Vis for life science applications
  • Pre-configured methods for nucleic acid and protein analysis
  • No warm up time needed—instant measurements from start up
  • USB connectivity to computers, printers and data storage devices
  • Use Smart Screen to design a power-up menu that displays frequently used tests
This spectrophotometer offers the flexibilty and performance a busy laboratory demands. The time-saving software allows you to measure DNA, RNA, and oligo concentrations. With the built-in Oligo calculator, determine molecular weight and theoretical Tm. Measure protein concentrations using Bradford, Lowry, BCA, and Biuret reagents. High-intensity xenon lamp and dual-beam optical geometry provide unsurpassed data quality.

USB connectivity allows easy access to USB memory devices. Data can be printed out using either a PCL printer or the optional internal printer.
What's Included:
Six-position cell holder, single-position cell holder, dust cover and US power cord. Printer model also includes one paper roll.
Product Type Spectrophotometer
Optics Dual-beam
Bandwidth 1.8 nm
Wavelength range 190 to 1100 nm
Wavelength accuracy ± 1.0 nm
Wavelength reproducibility ± 2.0 nm
Photometric range 0.3 to 125% T;
-0.1 A to 3 A
±9999 C
Photometric readout Transmittance, Absorbance, Concentration
Photometric noise <0.00030 @ 0 A;
<0.00055 @ 1.0 A;
<0.00085 @ 2.0 A
Photometric drift <0.0005 A/hour
Detector Dual silicon photodiodes
Output 1 USB Type A for USB external printer;
1 USB Type B for PC interface
Beam Type Double
Stray light <0.08% T @ 220;
340 nm (NaI, NaNO2);
<1.0% 198 to 200 nm (KCl)
Source lamp Xenon
Lamp life 5 years
Display Graphical 320 x 240 pixel, Backlit LCD, 3.8" x 2.8"
(9.6 x 7.1 cm)
Dimensions (in.) 11 3/4 W x 9 3/4 H x 15 3/4 D
Dimensions (cm) 30 W x 25 H x 40 D
CE Compliance Yes
Brand Thermo Scientific Genesys

Calibrations
Qty/Price
Availability

EACH
Usually ships in 44 days.

PACKAGE
Usually ships in 14 days or sooner.




Technical Resources
 
Related Products for Thermo Scientific GENESYS ™ 10S Bio UV/Visible Spectrophotometer, 6 cell
Image
  Product Detail
SKU

Thermo Scientific GENESYS 10S Bio UV/Vis Spectrophotometer; Printer/6 Cell

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Bio UV/Visible Scanning Spectrophotometers
Brand: Thermo Scientific Genesys


Peltier thermostated single cell holder for GENESYS 10 (222-238100)

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand: Thermo Scientific Genesys


GENESYS 10 nanoCell accessory, 0.2mm pathlength (222-227700)

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand: Thermo Scientific Genesys


GENESYS 10 nanoCell accessory, 1.0mm pathlength (222-227800)

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand: Thermo Scientific Genesys


GENESYS 10 nanoCell accessory, 0.2mm pathlength adapter (222-227900)

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand: Thermo Scientific Genesys


GENESYS 10 nanoCell accessory, 1.0mm pathlength adapter (222-228000)

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand: Thermo Scientific Genesys


GENESYS 10 nanoCell accessory with 0.2mm and 1.0mm pathlength adapter (222-230300)

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand: Thermo Scientific Genesys


Thermo Scientific GENESYS 10S UV/Vis Spectrophotometer; 1 & 6 Cell

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Visible and UV/Visible Scanning Spectrophotometers
Brand: Thermo Scientific Genesys


Thermo Scientific GENESYS 10S UV/Vis Spectrophotometer; Printer/1 & 6 Cell

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Visible and UV/Visible Scanning Spectrophotometers
Brand: Thermo Scientific Genesys


Thermo Scientific GENESYS 10S Visible Spectrophotometer; 1 & 6 Cell

Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Visible and UV/Visible Scanning Spectrophotometers
Brand: Thermo Scientific Genesys


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

Thermo Scientific GENESYS ™ 10S Bio UV/Visible Spectrophotometer, 6 cell
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Thermo Scientific GENESYS 10S Visible Spectrophotometer; Printer/1 & 6 Cell  |  Next: Thermo Scientific Glass Automatic Temperature Compensation Probe