Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-75800-59
Representative Photo Only. Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 153 GPM, intermittent duty, 15 HP, 6
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-75800-59
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Positive Displacement Pumps
Share
|
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 153 GPM, i
YO-75800-59
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Multiple designs—either dry pump cavity or lubricant filled—to best fit your system requirements
Complete isolation of pumped fluids
Self-priming and runs dry
Wide range of pump configurations and flows
No cups, packing, or seals for low maintenance
Will not break up delicate emulsions or create excessive frothing
The versatile VECTOR peristaltic pump series gives you pumping options you can’t get from standard positive displacement or centrifugal industrial pumps. The VECTOR pump is designed
to handle the high-viscosity fluids commonly found in industrial processing. Whether these fluids are pasty, pulpy or just plain thick, the VECTOR peristaltic pump can reliably start, stop and continuously pump such fluids at a wide range of pressures and flows. These pumps are ideal for abrasive fluids,
dyes, thick fluids, solids up 2.5 inches, shear-sensitive fluids, stringy material and corrosive fluids.
Specifications
Product Type
Positive Displacement Pump
Viscosity (Max)
100,000 cp
Liquid temperature (Max)
180°F (82°C)
Suction lift
22 ft (6.7 m)
Discharge (pressure)
116 psi (8 bar) max
Connections
6" 316 SS ANSI 150-lb flange
Max flow rate (GPM)
153
Max flow rate (Liters/min)
579.1
Max solid size
2-1/2" (64 mm)
Displacement (gal./rev)
5.28
Displacement (L/rev)
19.9
Series
2010
Manufacturer number
2010-HS-DD-M2
Model
2010-HS-DD-M2
Technical Resources
Related technical articles (Roll over for summary)
Positive Displacement Pump Types
How to Read a Pump Curve
Guide to Liquid Pumps
Show All...
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Related Searches for Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 153 GPM, i
Hypalon Large-Hose
Large-Hose
Cole-Parmer Large-Hose
Cole Large-Hose Peristaltic Pumps
Rubber Hose Peristaltic Pump
Dry Pumps
Hose Peristaltic
Norprene Pumps - Metering Pumps
Transfer Pumps Centrifugal Pumps
Replacement Hoses for Large-Hose Peristaltic Pumps
Related Products for Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 153 GPM, i
Image
Product Detail
SKU
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 57 GPM, co
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Hypalon
Model Number:
2009-HF-BB-D2
YO-75800-44
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 79 GPM, co
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Hypalon
Model Number:
2010-HS-DD-F2
YO-75800-54
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 39 GPM, in
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Hypalon
Model Number:
2008-HF-BB-J2
YO-75800-39
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 97 GPM, in
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Hypalon
Model Number:
2009-HF-BB-J2
YO-75800-49
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 23 GPM, co
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Hypalon
Model Number:
2008-HF-BB-D2
YO-75800-34
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 153 GPM, inte
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2010-ES-DD-M2
YO-75800-58
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 57 GPM, conti
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-EF-BB-D2
YO-75800-43
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 97 GPM, inter
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-EF-BB-J2
YO-75800-48
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 39 GPM, inter
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2008-EF-BB-J2
YO-75800-38
More Details
Large-Hose Peristaltic Pump, Dry Pump Cavity, EPDM Hose, 79 GPM, conti
Category: Positive Displacement Pumps
Description: Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2010-ES-DD-F2
YO-75800-53
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 153 GPM, i
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Large-Hose Peristaltic Pump, Dry Pump Cavity, Heavy Duty Natural Rubbe (2010-MS-DD-M2)
|
Next: Large-Hose Peristaltic Pump, Dry Pump Cavity, Hypalon Hose, 23 GPM, co
31125329-p-content_2 31125329pcontent2