Cole-Parmer – your technical experts in fluid handling, electrochemistry, and laboratory equipment.

 
Quick Order Form
 
 
More Items Add to Cart
Subscribe Today
Get the latest news, products,
special offers and more.
 
 
Masterflex Tubing & Pumps
Masterflex C/L Tubing Pumps
Masterflex L/S Tubing Pumps
Masterflex I/P Tubing Pumps
Masterflex  B/T  Tubing Pumps
 
pH Meters & Water Quality Products
pH Meters, Ion Meters and ISFET Meters
Representative photo only Oakton PCSTestr 35 Waterproof Multiparameter pH conductivity TDS salinity pocket tester
Electrodes, Laboratory
Buffers, Standards, and Solutions
 
Lab Equipment
Digi-Sense Single Input Thermocouple Thermometer, Type K/J with NIST
Balances & Scales
Baths and Circulators
Centrifuges
 
Featured Products
pH Benchtop Meter Kit by Thermo Scientific
Thermo Scientific GENESYS 10S UV-Visible Spectrophotometer with USB
Colder Aseptiquik Connectors

Bath for Bath/Magnetic Stirrer Systems, 8 L, 220 VAC/50 Hz
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Bath for Bath/Magnetic Stirrer Systems, 8 L, 120 VAC/60 Hz  |  Next: Bath oil, 6 gallons (9726-L20)