Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
15 liter cell culture BioBundle
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-29206-52
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Applikon BioBundle Fermentation Systems
Share
|
15 liter cell culture BioBundle
YO-29206-52
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Economical, complete and ready to assemble
Control pH, temperature DO, foam/level and agitation 29206-44 through a simple ez-Control touchscreen panel
Affordable and complete - systems include everything you need for a wide range of applications
Easy - packaged for fast, trouble-free assembly, setup requires no special skills or tools
BioBundle fermentation systems are the fastest, easiest and most economical way to get your fermentation projects up and running. Complete and ready to assemble. Every BioBundle system is made of the finest materials available and built to GMP standards. All material in contact with the bioreactor vessel is constructed of either 316L stainless steel, EPDM, silicone rubber, PTFE or borosilicate glass.
BioBundle bioreactors are available in 1-liter, 3-liters, 7 liters and 15 liters for mammalian cell culture or microbial processes, including bacteria, yeast, insect, fungi and plant cells. The P100 motor includes a 6:1 gear reduction to provide higher torque capability. All units are 110 VAC; for 220 VAC models, call our Application Specialists for availability.
Specifications
Product Type
BioBundle Fermentation
Working volume (Liters)
12
Vessel volume (Liters)
15
Port size (mm)
10, 12, 27
Manufacturer number
ZC3BEZ1C15
Brand
BioBundle
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
Related technical articles (Roll over for summary)
Sterilized Disposable Bioprocess Consumables
Cell Culture Fermentation Systems Selection Guide
Cole-Parmer Introduces Enhancements to Life Science Offering
Show All...
Show Less...
Related Searches for 15 liter cell culture BioBundle
Cell Culture Kits
Cell Culture Bio Reactors
Cell Culture Water Bath
Smallest Cover Glasses Cell Cultures
Glass Cell Culture Spinner Flasks
Wheaton Cell Culture Spin Flasks
Cell Culture Supplies
Cellbind Cell Culture Dishes
Cole-Parmer Systems for Cell Culture Applications
Cultrecoat Glass Cell Culture Flasks
Related Products for 15 liter cell culture BioBundle
Image
Product Detail
SKU
1 liter cell culture BioBundle
Category: Applikon BioBundle Fermentation Systems
Description: Applikon BioBundle Fermentation Systems - Cell Culture BioBundle
Brand:
BioBundle
YO-29206-40
More Details
3 liter cell culture BioBundle
Category: Applikon BioBundle Fermentation Systems
Description: Applikon BioBundle Fermentation Systems - Cell Culture BioBundle
Brand:
BioBundle
YO-29206-44
More Details
7 liter cell culture BioBundle
Category: Applikon BioBundle Fermentation Systems
Description: Applikon BioBundle Fermentation Systems - Cell Culture BioBundle
Brand:
BioBundle
YO-29206-48
More Details
15 liter microbial culture BioBundle (YO-29206-54)
Category: Applikon BioBundle Fermentation Systems
Description: Applikon BioBundle Fermentation Systems - Microbial Culture BioBundle
Brand:
BioBundle
YO-29206-54
More Details
1 liter microbial culture BioBundle
Category: Applikon BioBundle Fermentation Systems
Description: Applikon BioBundle Fermentation Systems - Microbial Culture BioBundle
Brand:
BioBundle
YO-29206-42
More Details
3 liter microbial culture BioBundle
Category: Applikon BioBundle Fermentation Systems
Description: Applikon BioBundle Fermentation Systems - Microbial Culture BioBundle
Brand:
BioBundle
YO-29206-46
More Details
7 liter microbial culture BioBundle
Category: Applikon BioBundle Fermentation Systems
Description: Applikon BioBundle Fermentation Systems - Microbial Culture BioBundle
Brand:
BioBundle
YO-29206-50
More Details
Systems for cell culture applications, water-jacketed, 220 VAC (YO-29200-15)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-15
More Details
Stackable cell culture system, one 1000-mL media reservoir
Category: Stackable Culture Systems
Description: INTEGRA Biosciences CELLine CL 1000 15-mL Stackable Cell Culture Chamber
Brand:
CELLine
YO-13052-20
More Details
Systems for cell culture applications, 115 VAC (YO-29200-42)
Category: Cole-Parmer Modular Fermentation Start-Up Kits
Description: Systems for Cell Culture Applications
Brand:
Cole-Parmer
YO-29200-42
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
15 liter cell culture BioBundle
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: 15 Gallon tank for mechanical pumps
|
Next: 15 liter microbial culture BioBundle (YO-29206-54)
31125329-p-content_2 31125329pcontent2