Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
DrumQuik Pro Dip Tube for Jerry Cans 330 mm
Click to enlarge
More Product Details
Specifications
Technical Resources
Technical information
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-31608-24
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
DrumQuik Drum Adapter Systems
Share
|
DrumQuik Pro Dip Tube for Jerry Cans, 330 mm
YO-31608-24
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Closed-system design ensures the safety of your plant, the environment, and your employees—at all phases of chemical handling
Ideal for 5-gallon jerry cans, 55-gallon drums, and IBCs
A peristaltic pump AODD, solenoid metering pump, or compressed air line dispenses the fluid from the container through use of its suction lift
The DrumQuik PRO system offers a simplified solution to your chemical dispensing needs. This system maintains a closed system, protecting your facility, your operating environment, and your employees from the potential dangers of chemical drips, spills and escaping fumes. The simple connections, rugged construction, and chemical-resistant materials handle a wide variety of applications—including acids and caustics, agricultural liquids, cleaning agents and sterilants, paints and coatings, and food oils and syrups. The base components are a reusable quick-disconnect coupler and a single-use drum insert assembly; both of which are made from environmentally friendly, food-grade polyethylene (PE). Order a convenient dispensing kit which includes all required components, or customize your own system with individual components. The coupler body and plugs mate equally well with all drum inserts listed.
What's Included:
Kits include a 1⁄2" NPT(M) coupler body, 2" buttress drum insert, dip tube, liquid line fitting, vent check valve, and shipping plug.
Specifications
Product Type
Dispensing System
Material
Polyethylene
Brand
DrumQuik
Manufacturer number
DQPRO DT 0410
Model
DQPRODT0330
Technical Resources
Chemical Dispensing White Paper
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Closed System Technology Drives the Trend Toward Safer, More Cost-Efficient Chemical Dispe
Pipe Thread Types and Designations
What to Consider when Selecting Couplings for Fluid Systems Selecting Couplings for Fluid Systems
Show All...
Closed System Technology Drives the Trend Toward Safer, More Cost-Efficient Chemical Dispensing
Quick Couplings Selection Guide
Sterilizing Fittings: The Critical Factors
Fittings Selection Guide
How to Read a Pump Curve
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Related Searches for DrumQuik Pro Dip Tube for Jerry Cans, 330 mm
Drumquik Fittings for Tubing
Drumquik Pro Drum Dispensing System
Quick Disconnect Dip Tube
Drumquik Pro
Colder Products Dip Tube Dqmdtubepp35
Jerry Cans
Drumquik Pur
Pro Drum
Dips Hdpe Fittings
Dr 11 Dips Hdpe Fittings
Related Products for DrumQuik Pro Dip Tube for Jerry Cans, 330 mm
Image
Product Detail
SKU
DrumQuik Pro Dip Tube for Drums, 902 mm
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
Model Number:
DQPRODT0902
YO-31608-25
More Details
DrumQuik Pro Dip Tube for Drums, 990 mm
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
Model Number:
DQPRODT0990
YO-31608-26
More Details
Atago Pen-Pro Dip-Style Digital Refractometer (3730 (PEN-PRO))
Category: Digital Handheld Refractometers
Description: Atago Pen-Pro Dip-Style Digital Refractometer
Brand:
ATAGO
Model Number:
3730 (PEN-PRO)
YO-02940-75
More Details
DrumQuik Pro Coupler, 1/2" NPTM (DQPRO120208)
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
Model Number:
DQPRO120208
YO-31608-20
More Details
DrumQuik Pro Coupler, 3/4" BSPM (DQPRO120712)
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
Model Number:
DQPRO120712
YO-31608-21
More Details
DrumQuik Pro 2" Buttress Adapter (DQPRO202BUT)
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
Model Number:
DQPRO202BUT
YO-31608-22
More Details
DrumQuik Pro BCS Buttress Adapter (DQPRO2056X4)
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
Model Number:
DQPRO2056X4
YO-31608-23
More Details
DrumQuik Pro Adapter, 1/2" NPTF x 3/8" OD (YO-31608-30)
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
YO-31608-30
More Details
DrumQuik Pro Adapter, 1/2" NPTF x 1/2" OD (YO-31608-31)
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
YO-31608-31
More Details
DrumQuik Pro Adapter, 1/2" NPTF x 5/8" OD (YO-31608-32)
Category: DrumQuik Drum Adapter Systems
Description: DrumQuik PRO Drum Dispensing System
Brand:
DrumQuik
YO-31608-32
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
DrumQuik Pro Dip Tube for Jerry Cans, 330 mm
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: DrumQuik Pro Dip Tube for Drums, 990 mm
|
Next: DrumQuik Pro Drum Insert Plug (YO-31608-37)
31125329-p-content_2 31125329pcontent2