Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
INDSTRL HOSE PMP 79GPM NATRBR
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-75800-51
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Positive Displacement Pumps
Share
|
industrial Hose Pump, Natural Rubber Hose, 79 GPM, continuous duty, 10
YO-75800-51
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Manufacturer number
2010-MS-DD-F2
Model
2010-MS-DD-F2
Related Searches for industrial Hose Pump, Natural Rubber Hose, 79 GPM, continuous duty, 10
Natural Rubber Hosing
Hose Pumps Business & Industrial
Large Industrial Oil Pumps Pictures
Cole-Parmer Continuous-Duty Pumps
Rubber Hose for Pump
Industrial Rubber Hoses
Small Industrial Hose Pumps
Hose Pumps
Large Rubber Hose
Peristaltic Hose Pumps
Related Products for industrial Hose Pump, Natural Rubber Hose, 79 GPM, continuous duty, 10
Image
Product Detail
SKU
industrial Hose Pump, Natural Rubber Hose, 23 GPM, continuous duty, 5
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2008-NF-BB-D2
YO-75800-31
More Details
industrial Hose Pump, Natural Rubber Hose, 57 GPM, continuous duty, 7.
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-NF-BB-D2
YO-75800-41
More Details
industrial Hose Pump, Natural Rubber Hose, 3.1 GPM, continuous duty, 1
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
3005-NF-PV-F2
YO-75800-61
More Details
industrial Hose Pump, Natural Rubber Hose, 153 GPM, intermittent duty,
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2010-MS-DD-M2
YO-75800-56
More Details
industrial Hose Pump, Natural Rubber Hose, 39 GPM, intermittent duty,
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2008-NF-BB-J2
YO-75800-36
More Details
industrial Hose Pump, Natural Rubber Hose, 97 GPM, intermittent duty,
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-NF-BB-J2
YO-75800-46
More Details
industrial Hose Pump, Natural Rubber Hose, 8.2 GPM, intermittent duty,
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
3005-NF-PV-J2
YO-75800-66
More Details
industrial Hose Pump, Nitrile Rubber Hose, 3.1 GPM, continuous duty, 1
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
3005-CF-PV-F2
YO-75800-63
More Details
industrial Hose Pump, Nitrile Rubber Hose, 8.2 GPM, intermittent duty,
Category: Positive Displacement Pumps
Description: Large-Hose, Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
3005-CF-PV-J2
YO-75800-68
More Details
Hose Replacement for 75800-4* pumps, Natural Rubber, fiber braided, st
Category: Hose
Description: Replacement Hoses for Large-Hose Peristaltic Pumps
Brand:
Cole-Parmer
Model Number:
2009-108-2329
YO-75800-86
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
industrial Hose Pump, Natural Rubber Hose, 79 GPM, continuous duty, 10
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: industrial Hose Pump, Natural Rubber Hose, 57 GPM, continuous duty, 7.
|
Next: industrial Hose Pump, Natural Rubber Hose, 8.2 GPM, intermittent duty,
31125329-p-content_2 31125329pcontent2