Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-06100-90
Representative photo only. Container sold separately. Replacement Cap for Collapsible Containers, PP, 38 mm
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-06100-90
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Collapsible Bottles
Share
|
Replacement Cap for Collapsible Containers, PP, 38 mm
YO-06100-90
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
38-mm replacement cap for collapsible containers.
Specifications
Product Type
Accessory
Related Searches for Replacement Cap for Collapsible Containers, PP, 38 mm
Cole-Parmer Hinged Lid Sample Containers
Cole-Parmer Collapsible Containers
Collapsible Basket
Shipping Cartons for Collapsible Containers
Cole-Parmer Graduated Sample Containers
Cole-Parmer Collapsible Basket
Graduated Sample Containers
Corning Snap-Seal Plastic Sample Containers
Collapsible Containers
Cole Collapsible Contain
Related Products for Replacement Cap for Collapsible Containers, PP, 38 mm
Image
Product Detail
SKU
Collapsible Basket, PP, Blue
Category: Plastic Storage Bins & Containers
Description: Collapsible Basket, PP
Brand:
Cole-Parmer
Model Number:
40003 BLUE
YO-06818-70
More Details
Collapsible Basket, PP, Red
Category: Plastic Storage Bins & Containers
Description: Collapsible Basket, PP
Brand:
Cole-Parmer
Model Number:
40002
YO-06818-72
More Details
Hinged-Lid Sample Containers, PP, 1/4 oz, 2500/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-00
More Details
Hinged-Lid Sample Containers, PP, 1/2 oz, 2000/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-10
More Details
Hinged-Lid Sample Containers, PP, 3/4 oz, 1500/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-20
More Details
Hinged-Lid Sample Containers, PP, 1 oz, 1300/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-30
More Details
Hinged-Lid Sample Containers, PP, 2 oz, 750/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-40
More Details
Hinged-Lid Sample Containers, PP, 4 oz, 300/pk
Category: Plastic Containers
Description: Hinged-Lid Sample Containers, PP
Brand:
Cole-Parmer
YO-66098-50
More Details
Lab-Tek Multi Purpose Lab Container, PP, 16 OZ, 120/CS
Category: Plastic Containers
Description: Lab-Tek Multi Purpose Lab Containers, PP
Brand:
Lab-Tek
Model Number:
4704
YO-06015-02
More Details
Lab-Tek Multi Purpose Lab Container, PP, 32 OZ, 120/CS
Category: Plastic Containers
Description: Lab-Tek Multi Purpose Lab Containers, PP
Brand:
Lab-Tek
Model Number:
4705
YO-06015-04
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Replacement Cap for Collapsible Containers, PP, 38 mm
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Replacement calibration standard, 80% ERH;Box of 5, for use with water (EA-80)
|
Next: Replacement cap for Kimax media bottles, PP, blue, 10/cs
31125329-p-content_2 31125329pcontent2