Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-68462-65
Surface-Mounting Bracket for Minihelic Gauges
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-68462-65
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Dwyer Magnehelic Differential Pressure Gauges
Share
|
Surface-Mounting Bracket for Minihelic Gauges
YO-68462-65
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Differential Pressure Gauge Accessories
Product Type
Differential Pressure Gauge Accessories
Brand
Dwyer
Brand
Dwyer
Model
A497
Model
A497
Manufacturer number
A-497
Manufacturer number
A-497
Technical Resources
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Related Searches for Surface-Mounting Bracket for Minihelic Gauges
Dwyer Back-Mount Direct Drive Pressure Gauges
For Mounting Bracket
Cole-Parmer Mounting Brackets
Shelf Mounting Bracket
Mounting Bracket Kit
Strobes - Surface-Mount
Mounting Brackets/Panels
Wall Mounting Bracket
Surface Mount Temperature Switches
Sensor Mounting Bracket
Related Products for Surface-Mounting Bracket for Minihelic Gauges
Image
Product Detail
SKU
Mounting bracket kit for Ion-X-Changer Filter Cartridges (YO-01506-60)
Category: Water Deionization Systems Accessories
Description: Mounting Brackets
Brand:
Cole-Parmer
YO-01506-60
More Details
Mounting Kit for Magnehelic Gauges
Category: Dwyer Magnehelic Differential Pressure Gauges
Description: Dwyer Pressure Gauge Accessories
Brand:
Dwyer
Model Number:
A-610
YO-68462-63
More Details
Air Filter Kit for Minihelic Gauges
Category: Dwyer Magnehelic Differential Pressure Gauges
Description: Dwyer Pressure Gauge Accessories
Brand:
Dwyer
Model Number:
A609
YO-68462-66
More Details
Universal Stirrer Bracket (916134)
Category: Laboratory Frame/Rod/Clamp Connectors
Description: Universal Stirrer Mounting Bracket
Brand:
Troemner
Model Number:
916134
YO-08041-40
More Details
Spriahelic 4 1/2" Direct Drive Pressure Gauge, 30 psi, 1/4" NPT(F) Bac
Category: Mechanical Pressure Gauges
Description: Back-Mount Direct Drive Pressure Gauges
Brand:
DWYER
Model Number:
7112-G030
YO-68063-60
More Details
Spriahelicreg> 4 1/2" Direct Drive Pressure Gauge, 60 psi,
Category: Mechanical Pressure Gauges
Description: Back-Mount Direct Drive Pressure Gauges
Brand:
DWYER
Model Number:
7112-G060
YO-68063-62
More Details
Spriahelicreg> 4 1/2" Direct Drive Pressure Gauge, 100 psi
Category: Mechanical Pressure Gauges
Description: Back-Mount Direct Drive Pressure Gauges
Brand:
DWYER
Model Number:
7112-G100
YO-68063-64
More Details
Spriahelicreg> 4 1/2" Direct Drive Pressure Gauge, 200 psi
Category: Mechanical Pressure Gauges
Description: Back-Mount Direct Drive Pressure Gauges
Brand:
DWYER
Model Number:
7112-G200
YO-68063-66
More Details
Spriahelicreg> 4 1/2" Direct Drive Pressure Gauge, 300 psi
Category: Mechanical Pressure Gauges
Description: Back-Mount Direct Drive Pressure Gauges
Brand:
DWYER
Model Number:
7112-G300
YO-68063-68
More Details
Spriahelicreg> 4 1/2" Direct Drive Pressure Gauge, 600 psi
Category: Mechanical Pressure Gauges
Description: Back-Mount Direct Drive Pressure Gauges
Brand:
DWYER
Model Number:
7112-G600
YO-68063-70
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Surface-Mounting Bracket for Minihelic Gauges
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Surface-Mount Refrigeration Alarm; temperature range; -25 - 30C, 110V (TA-2C-110)
|
Next: Surfactant (X+/X-) ISE Double junction solution (YO-27502-95)
31125329-p-content_2 31125329pcontent2