Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
YO-59004-03
Wastewater pH meter kit
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-59004-03
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
YO-17106-20
NIST-Traceable Calibration; pH Meter
$214.07 AUD/EACH/EACH
Usually ships in 23 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH/Ion Handheld Meters
Share
|
Hach SensION™+ Wastewater pH Meter Kit
YO-59004-03
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Basic all-in-one kits with everything you need to start testing
Guided menu navigation makes pH measurements fast and simple
Automatic buffer recognition at 2.00, 4.01, 7.00, 9.21 and 10.01
IP67-rated waterproof housing
The sensIon+ pH, mV and temperature meter is packaged with everything you need to start testing clean water or wastewater. Meters feature a backlit graphic LCD with easy to follow Icon-based user interface.
Other features include selectable auto-off, two point temperature re-adjustment, ORP calibration, a time measurement indicator for electrode life, and improved stabilization algorithm. The hard carrying case also acts as a workstation in the field.
Clean/Drinking water PH1 kit includes general pH/ATC electrode 59004-53.
Wastewater PH1 kit includes double junction pH/ATC electrode 59004-54.
What's Included:
pH electrode and temperature ATC probe, 4, 7, and 10 buffers 135-mL bottle, three AA batteries, and field carrying case.
Specifications
Product Type
Multiparameter Meters, Portable
Meter style
Portable
Dimensions (" W)
2.9375
Dimensions (" H)
1.5
Dimensions (" L)
7.3125
Range (pH)
0.00 to 14.00
Range (mV)
±1400
Range (Temperature °C)
-5 to 105
Range (Temperature °F)
23 to 221
Resolution (pH)
0.1/0.01
Resolution (Temperature)
0.1
Resolution (mV)
1
Accuracy (pH)
±0.02 pH
Accuracy (Temperature)
±0.3°C
Accuracy (mV)
±1.0%
Display
LCD
Calibration
Up to 2 points
Waterproof
Yes
Buffer recognition
USA, NIST
Power
Three AA batteries (included)
Battery life
500 hours
CE Compliance
Yes
Brand
Hach
Model
PH1 5051T
Accessories
Qty
/
Price
Availability
pH Electrode, Double-junction for Portable meter
YO-59004-52
EACH
Call for availability.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; pH Meter
YO-17106-20
EACH
Usually ships in 23 days.
Add to Cart
Technical Resources
sensION+ portable meter user manual (1497 KB)
Virtual Catalog (PDFs)
13-14 Catalog
13-14 International Catalog
More info
sensION+ portable meter user manual
Related technical articles (Roll over for summary)
pH FAQs
Testing the pH Value of Pure Water
Testing the pH value of Meat
Show All...
Testing the pH in Yogurt Production
Testing the pH value of Wine
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
pH Electrode Theory
Oakton® pH meter becomes a TV star!
Measuring the pH of Pure Water
pH Systems
pH Temperature Compensation
Temperature Compensation for pH Instruments
pH Measurement FAQs
Show Less...
Related Searches for Hach SensION™+ Wastewater pH Meter Kit
Wastewater Ph Meter
Online Wastewater Ph Meter
Hach Minilab Pocket Ph Meter
Hach Drinking Water Ph Meter
Ph Meters & Kits
Soil Ph Meter Kit Hanna
Beckman Ph Meter Kit
Student Digital Ph Meter Kit
Hach Sension Waterproof Ph/Mv Meter
Hach Ph Meter Hq Iid
Related Products for Hach SensION™+ Wastewater pH Meter Kit
Image
Product Detail
SKU
Hach SensION™+ Drinking Water pH Meter Kit
Category: pH/Ion Handheld Meters
Description: Hach sensION + Waterproof pH/mV Meter
Brand:
Hach
Model Number:
PH1 5050T
YO-59004-01
More Details
Hach SensION + Dissolved Oxygen Meter Kit
Category: pH/Ion Handheld Meters
Description: Hach sensION+ Waterproof Dissolved Oxygen Meter
Brand:
Hach
Model Number:
DO6 5020
YO-53013-12
More Details
Hach waterproof ISFET pH meter kit
Category: pH/Ion Handheld Meters
Description: Hach Waterproof ISFET Handheld pH/mV Meters
Brand:
Hach
Model Number:
IQ160-KIT
YO-99361-10
More Details
Hach SensION™+ Rugged Conductivity Meter Kit
Category: Conductivity/Resistivity/TDS Handheld Meters
Description: Hach sensION+ Waterproof Conductivity Meter
Brand:
HACH
Model Number:
EC5 5061
YO-19604-03
More Details
Hach SensION™+ Conductivity Meter Kit
Category: Conductivity/Resistivity/TDS Handheld Meters
Description: Hach sensION+ Waterproof Conductivity Meter
Brand:
HACH
Model Number:
EC5 5060
YO-19604-01
More Details
Hach waterproof ISFET pH meter only
Category: pH/Ion Handheld Meters
Description: Hach Waterproof ISFET Handheld pH/mV Meters
Brand:
Hach
Model Number:
IQ160NP
YO-99361-02
More Details
Thermo Scientific Orion 3-star benchtop pH meter kit, 115/220 VAC
Category: pH/Ion Benchtop Meters
Description: Thermo Scientific Orion 3-Star Benchtop pH Meters
Brand:
Thermo Scientific Orion
Model Number:
3-Star Meter
YO-58823-12
More Details
Thermo Scientific Orion 3-Star handheld pH meter kit
Category: pH/Ion Handheld Meters
Description: Thermo Scientific Orion 3-Star pH Meters
Brand:
Thermo Scientific Orion
Model Number:
3-Star
YO-58822-12
More Details
YSI EcoSense pH Meter Kit
Category: pH/Ion Handheld Meters
Description: YSI EcoSense pH100A pH/mV/Temp Meter
Brand:
YSI
Model Number:
PH100
YO-59351-34
More Details
Thermo Scientific Orion Star LogR pH Meter Kit with PerpHect glass-bod
Category: pH/Ion Benchtop Meters
Description: Thermo Scientific Orion Star LogR
™
pH and pH/Ion Meters
Brand:
Thermo Scientific Orion
YO-58822-62
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Hach SensION™+ Wastewater pH Meter Kit
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Hach SensION™+ Rugged Conductivity Meter Kit
|
Next: Hach StablCal 0.1 NTU Turbidity Standard, 500-mL
31125329-p-content_2 31125329pcontent2