Subtotal =  
Items =  
View Cart
Welcome
Home  >  L/S Easy-Load II Pump Heads   
Masterflex L/S Easy-Load II pump head for high-performance precision t (YO-77200-62)
YO-77200-62
Loading price...
Call for availability.
Qty
  • Automatic retention holds tubing securely in place with no manual adjustment—just load the tubing and start the pump
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation.
  • Improved occlusion bed geometry lengthens tubing life.
  • Polyphenylene sulfide (PPS) housing with either stainless steel (SS) or cold-rolled steel (CRS) rotor—choose CRS for economy or SS for greater durability and chemical resistance.
  • Model 77201-62 with adjustable occlusion gives you the option of loosening occlusion for longer tubing life or increasing occlusion for higher pressure.
  • Each pump head for high-performance precision pump tubing accepts tubing sizes L/S® 15, L/S® 24, L/S® 35, and L/S® 36. Achieve flow rates of 1.7 to 2900 mL/min (1 to 600 rpm).
These pump heads accept the four high-performance precision tubing sizes. High-performance precision tubing has a thicker wall than precision tubing enabling better pressure generation, greater suction lift, and greater ability to pump viscous fluids.
What's included: single-channel mounting hardware and a 15'' (38-cm) length of silicone pump tubing. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately.
Hot Tips:
With single or multiple channels, you can change tubing in each channel without removing the EASY-LOAD II pump head(s) from your drive.
Product Type Pump Head
mL/rev Depends on tubing selected.
Accepted tubing L/S® 15, L/S 24, L/S 35, L/S 36
Rotor material Stainless steel
Housing material Polyphenylene sulfide
Pressure Chart Click to View
Rollers Four; stainless steel
Occlusion Fixed
Dimensions 5"W x 4-3/4"H x 2-3/4"D
(12.7 x 12.1 x 7.0 cm)
Flow Chart Click to View
Automatic tubing retention Yes
Brand Masterflex
Manufacturer number 77200-62
Accessories
Qty/Price
Availability
SET of 1
Call for availability.
SET of 1
Call for availability.
SET of 1
Call for availability.
Upgraded Model
Qty/Price
Availability
EACH
Usually ships in 14 days or sooner.
Replacement parts
Qty/Price
Availability
SET of 1
Usually ships in 14 days or sooner.
Reconditioned & Like New
Qty/Price
Availability
RECOND 77200-62
EW-00151SM
at Call for Price
Usually ships in 14 days.
Technical Resources
Related Products for Masterflex L/S Easy-Load II pump head for high-performance precision t (YO-77200-62)
Image
  Product Detail
SKU
Masterflex L/S Easy-Load II pump head for high-performance precision t (YO-77200-52)
Category: L/S Easy-Load II Pump Heads
Description: L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
Brand: Masterflex
Masterflex L/S Easy-Load II pump head for high-performance precision t (YO-77201-62)
Category: L/S Easy-Load II Pump Heads
Description: L/S Easy-Load II Pump Heads for High-Performance Precision Tubing
Brand: Masterflex
Masterflex L/S Easy-Load 3 pump head for high-performance precision tu (YO-77800-62)
Category: L/S Easy-Load 3 Pump Heads
Description: L/S Easy-Load 3 Pump Heads for High-Performance Precision Tubing
Brand: Masterflex
Masterflex L/S Easy-Load 3 pump head for high-performance precision tu (YO-77800-52)
Category: L/S Easy-Load 3 Pump Heads
Description: L/S Easy-Load 3 Pump Heads for High-Performance Precision Tubing
Brand: Masterflex
Masterflex L/S Easy-Load II pump head for precision tubing, fixed occl (YO-77200-50)
Category: L/S Easy-Load II Pump Heads
Description: Masterflex L/S Easy-Load II Pump Heads for Precision Tubing
Brand: Masterflex
Masterflex L/S Easy-Load II pump head for precision tubing, fixed occl (YO-77200-60)
Category: L/S Easy-Load II Pump Heads
Description: Masterflex L/S Easy-Load II Pump Heads for Precision Tubing
Brand: Masterflex
Masterflex L/S Easy-Load II pump head for precision tubing, adjustable
Category: L/S Easy-Load II Pump Heads
Description: Masterflex L/S Easy-Load II Pump Heads for Precision Tubing
Brand: Masterflex
Masterflex L/S Easy-Load pump head for high-performance precision tubi (YO-07518-62)
Category: L/S Easy-Load Pump Heads
Description: L/S Easy-Load Pump Heads for High-Performance Precision Tubing
Brand: Masterflex
Replacement rotor assembly for Masterflex L/S Easy-Load II pump heads (YO-77200-58)
Category: L/S Easy-Load II Pump Head Parts
Description: Rotor Assemblies for L/S Easy-Load II Pump Heads
Brand: Masterflex
Replacement rotor assembly for Masterflex L/S Easy-Load II pump heads (YO-77200-59)
Category: L/S Easy-Load II Pump Head Parts
Description: Rotor Assemblies for L/S Easy-Load II Pump Heads
Brand: Masterflex
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call