Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Replacement plastic guard tip for 37950 52 RH temp probe
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-00082-FG
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
Replacement plastic guard/tip for 37950-52 RH/temp probe
YO-00082-FG
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Model
REPLACEMENT TIP
Related Searches for Replacement plastic guard/tip for 37950-52 RH/temp probe
Lab Products Replacements Accessories
I/P Replacements Accessories
Product Type Replace
Lab P Replacement Accessories
Lab Guard
Product Type Tips
Temp Probes
Type K Temp Probe
Medical Temperature Probe W Ecg Tips
Jackson Replacement Grinding Face Shields
Related Products for Replacement plastic guard/tip for 37950-52 RH/temp probe
Image
Product Detail
SKU
Type K 90°-angle surface thermocouple probe with guarded tip (TC/PR28875-3)
Category: Oakton Surface probes
Description: 90 Degree-Angle Surface Probes with Guarded Tip
Brand:
Oakton
Model Number:
TC/PR28875-3
YO-08445-10
More Details
Replacement Bulb guard for 12-mm electrodes. Pack of 4 guards.
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
BG50125-12C
YO-00070-KP
More Details
Replacement plastic storage box for Oakton Testrs
Category: General Accessories
Description: Replacement Accessories
Brand:
Oakton Instruments
Model Number:
56X239501-WP ROUND INSERT
YO-00093-qp
More Details
Replacement rpm cone tip (DTCA)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
DTCA
YO-87601-62
More Details
Type K Stainless steel straight guard surface thermocouple probe; 6.5" (112801E)
Category: Oakton Surface probes
Description: Surface Probes - Stainless Steel and Guarded Tip
Brand:
Oakton
Model Number:
112801E
YO-08447-21
More Details
Replacement RH/temperature probe
Category: General Accessories
Description: General Accessories and Replacement Items
Brand:
Cole-Parmer
YO-37850-05
More Details
Extech 45156 Replaceable Plastic Vane for Extech Model 45158 Anemomete
Category: General Accessories
Description: Replacement Accessories
Brand:
Extech
Model Number:
45156
YO-95001-98
More Details
Replacement blank glass plate (YO-28554-52)
Category: Thermo Scientific Owl Double-Sided Large-Vertical Electrophoresis Systems
Description: Accessories for Thermo Scientific Owl Double-Sided Large-Vertical Electrophoresis Systems
Brand:
Thermo Scientific
YO-28554-52
More Details
SHUR/Mark® Replacement marking pen tips for labeling slides or casset
Category: Slide Preparation Products
Description: SHUR/Mark Slide and Cassette Marking Pens
Brand:
Cole-Parmer
YO-48579-73
More Details
Rh/Temp Datalogger Kit w/ Graphing Software Custom Interface Cables (YO-10101-34)
Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand:
Davis
YO-10101-34
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Replacement plastic guard/tip for 37950-52 RH/temp probe
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Replacement pipettor battery (705500)
|
Next: Replacement plastic storage box for Oakton Testrs
31125329-p-content_2 31125329pcontent2