Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
AX-30 DISPOS SAMPLE PANS (100)
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-11110-55
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
AND Engineering Portable Balances
Share
|
A&D Disposable Sample Pans for Mositure Analyzers 100pcs.
YO-11110-55
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Weighing dishes
Related Searches for A&D Disposable Sample Pans for Mositure Analyzers 100pcs.
Disposable Weighing Pans
Disposable Aluminum Pans
Disposable Pans
Equipment Sampling
Pan Balance Accessories
Boxing Scales
Mettler Weighing Balance
Ohaus Balance and Scale Accessories
Ohaus Balance Moisture Anaylzer Accessories
Eqa Analyzer Sample Vial
Related Products for A&D Disposable Sample Pans for Mositure Analyzers 100pcs.
Image
Product Detail
SKU
A&D Washable Sample Pans for Mositure Analyzers 100pcs.
Category: AND Engineering Portable Balances
Description: A n D New Balances
Brand:
A&D
YO-11110-56
More Details
Disposable aluminum sample pans for Balances, pack of 250 (ALUMINUM PANS)
Category: Adam Equipment Balance Accessories
Description: Adam Equipment Balance Pans, Stands, and Bowls
Brand:
Adam Equipment
Model Number:
ALUMINUM PANS
YO-11700-66
More Details
ErgoClip ® accessory kit; includes 10 disposable aluminum pans, 50 we (11106707)
Category: Balances / Scales Accessories
Description: Mettler Toledo Balance Accessories
Brand:
ErgoClip
Model Number:
11106707
YO-11333-95
More Details
Ohaus Aluminum Sample Pans, 80/Box
Category: Ohaus Balance Accessories
Description: Ohaus Balance Moisture Anaylzer Accessories
Brand:
Ohaus
YO-11003-58
More Details
Ohaus Re-usable sample pans, 3/pk
Category: Ohaus Balance Accessories
Description: Ohaus Balance Moisture Anaylzer Accessories
Brand:
Ohaus
YO-29811-17
More Details
Round Sample Vials for i-LAB, 25-mm Diameter; Pack of 6
Category: i-LAB Handheld Analyzing Spectrometers
Description: Sampling Accessories and Adaptors for i-LAB Handheld Spectrometers
Brand:
i-LAB
YO-83080-72
More Details
Fyrite Tech Combustion Analyzer
Category: Combustion Analyzers
Description: Bacharach Fyrite Tech Combustion Analyzers
Brand:
Bacharach Fyrite
YO-86491-00
More Details
Fyrite Tech Combustion Analyzer With CO
Category: Combustion Analyzers
Description: Bacharach Fyrite Tech Combustion Analyzers
Brand:
Bacharach Fyrite
YO-86491-10
More Details
Drager CMS Analyzer
Category: Gas Detection-Tubes
Description: Drager CMS Analyzer
Brand:
Drager
YO-86513-00
More Details
Disposable bailer, PVC, 1-2/3" OD x 3' (BPWSU1.66X36)
Category: Disposable Bailers
Description: Polyvinylchloride (PVC) 1-2/3" OD Bailers
Brand:
Cole-Parmer
Model Number:
BPWSU1.66X36
YO-99154-11
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
A&D Disposable Sample Pans for Mositure Analyzers 100pcs.
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: A&D Compact Balance 400g x 0.01g Counting, Percentage, Comaparator Mod
|
Next: A&D Eki Portable Balance, 120 g x 0.01 g
31125329-p-content_2 31125329pcontent2