Extractor/concentrator

YO-34510-00
Loading price...

Call for availability.
Qty
    Accelerated One-Step™ modular extractor/concentrator performs extraction, concentration, and drying—all in one apparatus! Features PTFE hydrophobic membranes for speedy drying. Meets EPA and CLP standards. Includes extractor cup and body, Snyder sidearm, membrane assembly, and connectors. Order concentrator tube and condenser separately below to fit your specific application.

    Accessories
    Qty/Price
    Availability

    CASE of 1

    Call for availability.

    CASE of 1

    Call for availability.
    Allihn condenser
    YO-34510-03

    CASE of 1

    Call for availability.




    Technical Resources
     
    Related Products for Extractor/concentrator
    Image
      Product Detail
    SKU

    Concentrator tube, 100 mL (YO-34510-01)

    Category: Analytical Glassware
    Description: Accelerated One-Step Modular Extractor/Concentrator
    Brand: One-Step


    Jacketed concentrator tube, 100 mL (YO-34510-02)

    Category: Analytical Glassware
    Description: Accelerated One-Step Modular Extractor/Concentrator
    Brand: One-Step


    Allihn condenser (YO-34510-03)

    Category: Analytical Glassware
    Description: Accelerated One-Step Modular Extractor/Concentrator
    Brand: One-Step


    Kuderna-Danish Concentrator without springs, 500mL capacity, 615mm H

    Category: Kuderna-Danish Concentrator
    Description: Concentrator without springs
    Brand: Corning


    Kuderna-Danish Concentrator without springs, 1L capacity, 720mm H

    Category: Kuderna-Danish Concentrator
    Description: Concentrator without springs
    Brand: Corning


    Cole-Parmer Ion Concentration Controller

    Category: Ion Control
    Description: Cole-Parmer Ion Concentration Controller
    Brand: Cole-Parmer


    Cole-Parmer Control Guard FCS Chemical Concentration Controller

    Category: Diaphragm Pumps
    Description: Cole-Parmer Concentration Controller Systems
    Brand: Cole-Parmer


    Cole-Parmer Concentration Contoller Inductive Probe - Inductive Temperature Compensating Toroidal Probe wtih 25ft

    Category: Diaphragm Pumps
    Description: Cole-Parmer Concentration Controller Systems
    Brand: Cole-Parmer


    Cole-Parmer Concentration Contoller Conductive Probe - P-120S Conductive Probe

    Category: Diaphragm Pumps
    Description: Cole-Parmer Concentration Controller Systems
    Brand: Cole-Parmer


    Cole-Parmer Concentration Contoller-Air Solenoid Valve (mounted), 3/8" brass (110V)

    Category: Diaphragm Pumps
    Description: Cole-Parmer Concentration Controller Systems
    Brand: Cole-Parmer


    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
    Questions and Answers
    Product Reviews