Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Representative photo only
Replacement clear conductivity cover for 35630 50 probe
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
YO-00120-JP
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
General Accessories
Share
|
Replacement clear conductivity cover for 35630-50 probe
YO-00120-JP
Loading price...
Call for availability.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Model
15X278601
Related Searches for Replacement clear conductivity cover for 35630-50 probe
Conductivity Meter with Flow Thru Probe
Probe Cap
Reflective Wall Covering Lab Supply
Cover Roll Strech Lab Supplies
Ph Accessories
Product Type Crucibles
Crucibles
Lab Products Crucibles
Water Conductivity Probe Sensor
Cole-Parmer Threaded Probes with Replaceable Tip
Related Products for Replacement clear conductivity cover for 35630-50 probe
Image
Product Detail
SKU
Replacement pH cap for 35630-50 probe
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
Model Number:
15X278301
YO-00120-JQ
More Details
Replacement Quartz Crucible Cover, fits 34526-50 and -52. Case of 1
Category: Crucibles
Description: Quartz Crucible Cover Replacement
Brand:
Lab Products
Model Number:
319012-0050
YO-34526-54
More Details
Replacement pH/Conductivity/TDS Sensor Module (YO-35425-50)
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Oakton Instruments
YO-35425-50
More Details
Replacement frits, 10/pack (YO-02010-50)
Category: General Accessories
Description: Replacement Accessories
Brand:
Lab Products
YO-02010-50
More Details
Replacement clear yellow sleeve tube for 35608-50 & 35606-55, Ultem
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Lab Products
Model Number:
15X245501
YO-00162-SQ
More Details
Replacement motor, explosion-proof, 230 VAC, 50 Hz
Category: Cole-Parmer Drum Pump Motors
Description: Replacement Motors
Brand:
Cole-Parmer
Model Number:
SP-400-2
YO-70604-23
More Details
Clear C-FLEX Tubing, 1/50"ID x 1/12"OD, 25 ft/pack
Category: Flexible Tubing
Description: Clear C-FLEX Tubing
Brand:
C-FLEX
YO-06422-00
More Details
Eutech Instruments COND 1000 Conductivity Controller, 110 VAC, 50/60 H
Category: 1/4-DIN Conductivity Control
Description: Eutech Instruments COND 1000 Conductivity Controllers
Brand:
Eutech Instruments
YO-19506-00
More Details
Eutech Instruments COND 1000 Conductivity Controller, 220 VAC, 50/60 H
Category: 1/4-DIN Conductivity Control
Description: Eutech Instruments COND 1000 Conductivity Controllers
Brand:
Eutech Instruments
YO-19506-05
More Details
Replacement platinum conductivity cell
Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand:
Extech
Model Number:
EC405
YO-53026-13
More Details
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Replacement clear conductivity cover for 35630-50 probe
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
Previous: Replacement Classifier Lid (500 051 00)
|
Next: Replacement clear yellow sleeve tube for 35608-50 & 35606-55, Ultem
31125329-p-content_2 31125329pcontent2