Lubricant Super Lube (1cc) For Use with Cole-Parmer's Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788316) (YO-74905-86)

YO-74905-86
Loading price...

Call for availability.
Qty
Model 788316





Technical Resources
 
Related Products for Lubricant Super Lube (1cc) For Use with Cole-Parmer's Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788316) (YO-74905-86)
Image
  Product Detail
SKU

Lubricant Super Lube (1cc) For Use with Cole-Parmer's Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788316) (YO-74903-86)

Category: ColeParmer® Touch Screen Syringe Pumps
Description: Cole-Parmer® Touch Screen Syringe Pump Accessories
Brand: Cole-Parmer


5 Meter Communication Cable (USB) for Pump to Pump use with Cole-Parmer Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788307) (YO-74905-76)

Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand: Cole-Parmer


Adapter D Sub to terminal Block For Use with Cole-Parmer's Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788312) (YO-74905-82)

Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand: Cole-Parmer


Footswitch with Phono Jack Plug For Use with Cole-Parmer's Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (780225) (YO-74905-83)

Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand: Cole-Parmer


Hex Key For Use with Cole-Parmer's Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788315) (YO-74905-85)

Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand: Cole-Parmer


2 Meter Communication Cable (RS485) for Pump to Pump use with Cole-Parmer Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788304) (YO-74905-73)

Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand: Cole-Parmer


2 Meter Communication Cable (USB) for Pump to Pump use with Cole-Parmer Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788306) (YO-74905-75)

Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand: Cole-Parmer


Line Cord (US - 115V) for use with Cole-Parmer Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788308) (YO-74905-78)

Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand: Cole-Parmer


Adapter Digital I/O (15 Pin to 9 Pin) for use with Cole-Parmer Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788312) (YO-74905-81)

Category: Syringes and Accessories for Syringe Pumps
Description: Cole-Parmer Touch Screen Syringe Pumps Accessories
Brand: Cole-Parmer


Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Product Reviews

Lubricant Super Lube (1cc) For Use with Cole-Parmer's Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788316) (YO-74905-86)
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers

Previous: Lubricant Super Lube (1cc) For Use with Cole-Parmer's Syringe Pump, Infusion and Withdrawal design, Touchscreen Control (788316) (YO-74903-86)  |  Next: Lucite plunger for DO probe 05520-13