Adam Equipment Anti-vibration Table
YO-11120-92
Loading price...
Call for availability.
Qty
Product Type Antivibration Platform
Auto shutoff Y
Calibration External
Display Backlit LCD
Battery Rechargeable
Low battery indicator Y
RS-232 interface Y
Brand Adam Equipment
Model 104008036
Technical Resources
Related Products for Adam Equipment Anti-vibration Table
Image
  Product Detail
SKU
Adam Equipment Security lock and cable
Category: Adam Equipment Balance Accessories
Description: Adam Equipment Anti-theft Devices
Brand: Adam Equipment
Adam Equipment Kensington type security lock and cable
Category: Adam Equipment Balance Accessories
Description: Adam Equipment Anti-theft Devices
Brand: Adam Equipment
Mat & Table Top Cleaner, 1 Quart spray bottle
Category: ESD Solutions
Description: Mat & Table Top Cleaner
Brand: Cole-Parmer
ACL Staticide 6002, Mat and Table Top Cleaner, 1 Gallon
Category: ESD Solutions
Description: Mat & Table Top Cleaner
Brand: Staticide
Anti-static polypropylene weighing funnel, micro (YO-06127-00)
Category: Weighing Funnels
Description: Anti-Static Weighing Funnels, PP
Brand: Anti-Static
Anti-static polypropylene weighing funnel, small (YO-06127-02)
Category: Weighing Funnels
Description: Anti-Static Weighing Funnels, PP
Brand: Anti-Static
Anti-static polypropylene weighing funnel, medium (YO-06127-04)
Category: Weighing Funnels
Description: Anti-Static Weighing Funnels, PP
Brand: Anti-Static
Anti-static polypropylene weighing funnel, large (YO-06127-06)
Category: Weighing Funnels
Description: Anti-Static Weighing Funnels, PP
Brand: Anti-Static
Anti-static polypropylene weighing funnel, extra large (YO-06127-08)
Category: Weighing Funnels
Description: Anti-Static Weighing Funnels, PP
Brand: Anti-Static
Tri-Flow Food-Grade Anti-Seize Thread Compound, 9-oz jar
Category: Lubricants
Description: Anti-Seize Compounds
Brand: Tri-Flow
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Do you have questions about this product?
get answers from real customers and in-house experts with AnswerBox.
1 Question | 1 Answer
Displaying question 1
  • Q:
    I would like to know the size (WxLxH), thickness, and type of vibration isolation used for this product.
    Asked on 7/23/2013 by Ionut from Houston, TX
    1 answer
    • STAFF EXPERT
      A:
      The working surface of of the table is 400mm x 450mm; the overall dimensions are 800mm x 600mm x 782mm (W x D x H). The table comprises two separate parts: 1) an internal working surface consisting of a large granite slab mounted on adjustable rubber shock absorbing mounts, and; 2) an external service table with aluminium frame and adjustable leveling feet. Both components help to eliminate surrounding vibration.
      Answered on 7/23/2013 by James from Technical Support
Displaying question 1
Do you have a question about this product? 
Product Reviews