CHEMets Colorimetric Phenol Test Kit
YO-00290-30
Loading price...
Usually ships in 14 days.
Qty
  • CHEMets test kits are portable and self-contained
  • Vacuum sealed reagents and handy comparator chart for quick, visual results
CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely (see footnotes).
What's included: a polypropylene case with instructions, all necessary apparatus, and reagent vials for 30 tests (except detergent test kit, which has 20 tests). Refill kits contain 30 vials each (detergent refill kit has 20 vials).
Product Type Water Quality Test Kits
Range 0 to 1, 0 to 12 ppm
Test Phenol
Brand CHEMets
Model K-8012
Manufacturer number K-8012
Accessories
Qty/Price
Availability
BOX of 30
Usually ships in 14 days.
Technical Resources
Related Products for CHEMets Colorimetric Phenol Test Kit
Image
  Product Detail
SKU
CHEMets Colorimetric Copper Test Kit
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Iron Test Kit
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Detergent Test Kit (K-9400)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Sufide Test Kit (K-9510)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Phosphate Test Kit (K-8510)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Silica Test Kit (K-9010)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Phosphate, HR, Test Kit
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Ammonia/nitrogen Test Kit
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Chlorine DDPD Test Kit (K-2505)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
CHEMets Colorimetric Hydrogen Peroxide Test Kit (K-5510)
Category: Titrets and CHEMets Test Kits
Description: CHEMets Colorimetric Test Kits
Brand: CHEMets
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call
Questions and Answers
Do you have questions about this product?
get answers from real customers and in-house experts with AnswerBox.
1 Question | 1 Answer
Displaying question 1
  • Q:
    Would this product register the presence of tert-Butylphenol if the measurement range was accurate?
    Asked on 11/30/2011 by Jay from St Louis, MO
    1 answer
    • STAFF EXPERT
      A:
      We will need to know if the tert-butylphenol is o-tert-butyl, m-tert-butyl, or p-tert-butyl (or 2-, 3- or 4-tert-butylphenol). If it is p- or 4-, this kit will not be able to measure it. If it's o- or m- (2- or 3-), the kit will probably measure it but it is not confirmed how sensitive the kit will be, as this kit has not been tested experimentally with tert-butylphenols.
      Answered on 11/30/2011 by Rita from Customer Service
Displaying question 1
Do you have a question about this product? 
Product Reviews
Customers Who Bought This Item Also Bought
CHEMets Colorimetric Phenol Test Kit
Popularity |  Our Choices |  All-Around Favorites |  Title |  Product Category |  Recommended
Popularity |  Favorites |  Title |  Price |  Part Number |  Category
Title |  Brand |  Category |  Price |  Popularity |  Best Sellers
Previous: CHEMets Colorimetric Iron Test Kit  |  Next: CHEMets Colorimetric Phosphate Test Kit (K-8510)