Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 10

Abc Vial™ Convenience Kits with Amber Vial and Ptfe   to   Ac Adapter

Image
  Product Detail
SKU

ABC Vial™ convenience kits with amber vial and PTFE/red silicone se

Category: Wheaton 12 X 32 mm Autosampler Vials
Description: WHEATON ABC 12 X 32 MM AUTOSAMPLER VIALS 100/PK
Brand: Wheaton
Model Number: W225157-01


ABC Vial™ convenience kits with amber with write-on patch vial and (W225153-01)

Category: Wheaton 12 X 32 mm Autosampler Vials
Description: WHEATON ABC 12 X 32 MM AUTOSAMPLER VIALS 100/PK
Brand: Wheaton
Model Number: W225153-01


ABC Vial™ convenience kits with clear vial and PTFE/red silicone se

Category: Wheaton 12 X 32 mm Autosampler Vials
Description: WHEATON ABC 12 X 32 MM AUTOSAMPLER VIALS 100/PK
Brand: Wheaton
Model Number: W225154-01


ABC Vial™ convenience kits with clear vial and PTFE/silicone septa (W225150-04)

Category: Wheaton 12 X 32 mm Autosampler Vials
Description: WHEATON ABC 12 X 32 MM AUTOSAMPLER VIALS 100/PK
Brand: Wheaton
Model Number: W225150-04


ABC Vial™ convenience kits with clear vial and PTFE/silicone/PTFE s (W225150-03)

Category: Wheaton 12 X 32 mm Autosampler Vials
Description: WHEATON ABC 12 X 32 MM AUTOSAMPLER VIALS 100/PK
Brand: Wheaton
Model Number: W225150-03


ABC Vial™ convenience kits, clear with write-on patch vial and PTFE

Category: Wheaton 12 X 32 mm Autosampler Vials
Description: WHEATON ABC 12 X 32 MM AUTOSAMPLER VIALS 100/PK
Brand: Wheaton
Model Number: W225155-01


Absolute Pressure Module 14,500 psi

Category: Cole-Parmer Process Recorders
Description: Multifunction Portable Calibrators
Brand: Palmer Wahl


Absolute Pressure Module 14.5 psi

Category: Cole-Parmer Process Recorders
Description: Multifunction Portable Calibrators
Brand: Palmer Wahl


Absolute Pressure Module 145 psi

Category: Cole-Parmer Process Recorders
Description: Multifunction Portable Calibrators
Brand: Palmer Wahl


Absolute Pressure Module 1450 psi

Category: Cole-Parmer Process Recorders
Description: Multifunction Portable Calibrators
Brand: Palmer Wahl


Absolute Pressure Module 43.5 psi

Category: Cole-Parmer Process Recorders
Description: Multifunction Portable Calibrators
Brand: Palmer Wahl


Absolute Pressure Module 435 psi

Category: Cole-Parmer Process Recorders
Description: Multifunction Portable Calibrators
Brand: Palmer Wahl


Absolute Pressure Module 4350 psi

Category: Cole-Parmer Process Recorders
Description: Multifunction Portable Calibrators
Brand: Palmer Wahl


Absolute pressure monitor, 0 to 10 psia

Category: Pressure Transmitters and Pressure Transducers
Description: Setra Digital Barometric and Absolute Pressure Monitor
Brand: Setra
Model Number: 370 0 TO 10 PSIA


Absolute pressure monitor, 0 to 100 psia

Category: Pressure Transmitters and Pressure Transducers
Description: Setra Digital Barometric and Absolute Pressure Monitor
Brand: Setra
Model Number: 370 0 TO 100 PSIA


Absolute pressure monitor, 0 to 20 psia

Category: Pressure Transmitters and Pressure Transducers
Description: Setra Digital Barometric and Absolute Pressure Monitor
Brand: Setra
Model Number: 370 0 TO 20 PSIA


Absolute pressure monitor, 0 to 50 psia

Category: Pressure Transmitters and Pressure Transducers
Description: Setra Digital Barometric and Absolute Pressure Monitor
Brand: Setra
Model Number: 370 0 TO 50 PSIA


Absorbent Zorb Sheet™, 100/cs

Category: Spill Cleanup
Description: Absorbent Zorb Sheet
Brand: Zorb Sheet
Model Number: 44212


Absorptive Silencer for Compressors or Vacuum Pumps, 155 SCFM

Category: Accessories for Compressors - Air Equipment
Description: Absorptive Silencers
Brand: Cole-Parmer
Model Number: SLCR150


Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM

Category: Accessories for Compressors - Air Equipment
Description: Absorptive Silencers
Brand: Cole-Parmer
Model Number: SLCR100


Absorptive Silencer for Compressors or Vacuum Pumps, 55 SCFM

Category: Accessories for Compressors - Air Equipment
Description: Absorptive Silencers
Brand: Cole-Parmer
Model Number: SLCR125


AC Adapter 110V (705352)

Category: General Accessories
Description: Replacement Accessories
Brand: Lab Products
Model Number: 705352


AC adapter for 65530-16, -18, 20, and -64

Category: General Accessories
Description: Replacement Accessories
Brand: Lab Products
Model Number: 40077


Ac Adapter For Datalogging Manometers 68603-01, -03, -05

Category: Digital and Analog Manometers
Description: Datalogging Manometers
Brand: HVAC/R
Model Number: 840096


AC Adapter for SPER 850008 Light Meter (YO-01588-85)

Category: Datalogging Light Meters
Description: Datalogging Light Meter
Brand: Cole-Parmer


AC adapter for Temperature/Humidity Dataloggers with Digital Display (R157)

Category: General Accessories
Description: Replacement Accessories
Brand: Lab Products
Model Number: R157


AC adapter, 100 to 240VAC (1754)

Category: pH/Ion Meter Accessories
Description: Water Analysis Accessories
Brand: Lab Products
Model Number: 1754


AC adapter, 115 VAC

Category: Force Gauge Accessories
Description: AC Adapter for Compact Digital Force Gauge
Brand: Mecmesin


AC adapter, 115 VAC, 5-ft cable with U.S. plug (YO-08406-50)

Category: General Accessories
Description: Replacement Accessories
Brand: Cole-Parmer




Abc Vial™ Convenience Kits with Amber Vial and Ptfe to Ac Adapter

>