Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 16

Ac,   to   Accu-Cal Digital Test Gauge

Image
  Product Detail
SKU

Ac, With Condensate Evaporator,16.3" X 23.4" X 18.7", 6.8amps, 230V (YO-30002-30)

Category: Wall Mount Enclosures, Cabinets, & Racks
Description: Enclosure Air Conditioners
Brand: Rittal


Ac, With Condensate Evaporator,16.3" X 23.4" X 18.7", 6amps, 115V (YO-30002-27)

Category: Wall Mount Enclosures, Cabinets, & Racks
Description: Enclosure Air Conditioners
Brand: Rittal


Ac, With Condensate Evaporator,16.3" X 23.4" X 18.7", 8.7amps, 115V (YO-30002-29)

Category: Wall Mount Enclosures, Cabinets, & Racks
Description: Enclosure Air Conditioners
Brand: Rittal


AC/DC Voltage Tester

Category: Voltage & Current Testers/Detectors
Description: Extech Receptacle Testers
Brand: Extech
Model Number: ET25 10/PK


AC1008 : Table Top Mounting Kit for BGI

Category: Force Gauges
Description: Mark-10 Universal Force and Torque Gauge and Sensors
Brand: Mark-10
Model Number: AC1008


AC168640050XYDENZALDEHYDE 5G (YO-88170-83)

Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand: Cole-Parmer


AC243250250CARBONITRILE 10ML (YO-88274-11)

Category: Chemicals, Reagents, & Lubricants
Description: Specialty Chemicals
Brand: Cole-Parmer


Ac504:Carrying Case (YO-10104-49)

Category: Air Sampling and Gas Analysis
Description: Air Sampling and Gas Analysis Accessories
Brand: Davis


Academic Starter Kit for Nicolet iS5 FT-IR Spectrometer

Category: Infrared (IR) Spectroscopy
Description: Starter Kits and Accessories for Thermo Scientific NICOLET IR100 FT-IR Spectrometer
Brand: Thermo Scientific


Accelerometer, vibration sensor with integral cable and 1/4-28 tap (6400-031)

Category: General Accessories
Description: Replacement Accessories
Brand: Lab Products
Model Number: 6400-031


Accessories - 200 Lbf Capacity - 1.0 Lbf Resolution

Category: Force Gauges
Description: Ametek High Capacity Mechanical force Gauges
Brand: Ametek
Model Number: DG200


Accessory burner ring kit for burner stand 3631050 (AK-2001)

Category: General Accessories
Description: Replacements Accessories
Brand: Lab Products
Model Number: AK-2001


Accessory Key Pack for Access Control Option, for UxF.

Category: Freezer Accessories
Description: Thermo Scientific Revco Accessories for Upright and Chest Ultra-Low (-86°C) Temperature Freezers
Brand: Thermo Scientific


Accessory Kit for Pressure and Air Velocity Measurment (YO-10400-50)

Category: General Accessories
Description: General Accessories and Replacement Items
Brand: MODUS


Accessory Shelf, 2-ft (YO-09109-80)

Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand: Labconco


Accessory Shelf, 3-ft (YO-09109-81)

Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand: Labconco


Accessory Shelf, 4-ft (YO-09109-82)

Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand: Labconco


Accessory Shelf, 5-ft (YO-09109-83)

Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand: Labconco


Accessory Shelf, 6-ft (YO-09109-84)

Category: Balance Enclosures
Description: Labconco Filtered Balance Systems
Brand: Labconco


Accessory Tray for Multi-Microplate Genie

Category: Shaker Accessories
Description: Accessory Tray for Multi-Microplate Genie
Brand: Scientific Industries
Model Number: SI-4010


Accordian sleeves; 24"L with hands (5005900)

Category: Sleeves with Interchangeable Hands
Description: Glove Box Sleeves
Brand: Cole-Parmer
Model Number: 5005900


ACCRA LAB Gram stain set, stabilized

Category: Microbiology
Description: ACCRA lab gram stains
Brand: Accra Lab


AccTech Cleaning Pads, Alco-Pad

Category: Surface Cleaners
Description: AccTech Cleaning Pads
Brand: Alco-Pad
Model Number: CT806


AccTech Cleaning Pads, AllPurpose Cleaner

Category: Surface Cleaners
Description: AccTech Cleaning Pads
Brand: AccTech
Model Number: CT9401


AccTech Cleaning Pads, OpticPad

Category: Surface Cleaners
Description: AccTech Cleaning Pads
Brand: OpticPad
Model Number: CT811


AccTech Cleaning Pads, TexStat® (CT814)

Category: Surface Cleaners
Description: AccTech Cleaning Pads
Brand: TexStat
Model Number: CT814


Accu-cal Digital Test Gauge, Vac To 10,000 Psi

Category: Digital Pressure Gauges and Meters
Description: Accu-Cal Plus Digital Test Gauge
Brand: Accu-Cal
Model Number: 75514-38B55


Accu-cal Digital Test Gauge, Vac To 10,000 Psi, W/ Flange

Category: Digital Pressure Gauges and Meters
Description: Accu-Cal Plus Digital Test Gauge
Brand: Accu-Cal
Model Number: 75514-38B15


Accu-cal Digital Test Gauge, Vac To 10,000 Psi, W/ Flange, Loop Powere

Category: Digital Pressure Gauges and Meters
Description: Accu-Cal Plus Digital Test Gauge
Brand: Accu-Cal
Model Number: 76514-38B15


Accu-cal Digital Test Gauge, Vac To 10,000, Loop Powered

Category: Digital Pressure Gauges and Meters
Description: Accu-Cal Plus Digital Test Gauge
Brand: Accu-Cal
Model Number: 76514-38B55




Ac, to Accu-Cal Digital Test Gauge

>